Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 77721380-77721605 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNAQ
Entrez Gene ID
2776
Gene Name
G protein subunit alpha q
Gene Aliases
CMC1, G-ALPHA-q, GAQ, SWS
UniGene
Hs.269782
Gene Chromosome Location
Chr.9: 77716274-78031449 on Build GRCh38
Transcripts
NM_002072.4 XM_017014628.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3658636 c.899G>A p.R300K Chr.9: 77721504-77721504 on Build GRCh38
3658635 c.1018G>C p.V340L Chr.9: 77721385-77721385 on Build GRCh38
6672195 c.1012C>T p.R338C Chr.9: 77721391-77721391 on Build GRCh38
123394 c.974A>T p.Y325F Chr.9: 77721429-77721429 on Build GRCh38
5004965 c.1013G>A p.R338H Chr.9: 77721390-77721390 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775693911 Chr.9: 77721503-77721503 on Build GRCh38
rs757656161 Chr.9: 77721556-77721556 on Build GRCh38
rs149624211 Chr.9: 77721569-77721569 on Build GRCh38
rs766188046 Chr.9: 77721519-77721519 on Build GRCh38
rs879087534 Chr.9: 77721577-77721577 on Build GRCh38
rs144945100 Chr.9: 77721479-77721479 on Build GRCh38
rs1025189626 Chr.9: 77721543-77721543 on Build GRCh38
rs183713546 Chr.9: 77721446-77721446 on Build GRCh38
rs113500437 Chr.9: 77721559-77721559 on Build GRCh38
rs774951302 Chr.9: 77721496-77721496 on Build GRCh38
rs936701973 Chr.9: 77721530-77721530 on Build GRCh38
rs755652968 Chr.9: 77721548-77721548 on Build GRCh38
rs554382469 Chr.9: 77721538-77721538 on Build GRCh38
rs376987002 Chr.9: 77721493-77721493 on Build GRCh38
rs760869735 Chr.9: 77721504-77721504 on Build GRCh38
rs751516475 Chr.9: 77721452-77721452 on Build GRCh38
rs374303316 Chr.9: 77721506-77721506 on Build GRCh38
rs748971807 Chr.9: 77721545-77721545 on Build GRCh38
rs776922380 Chr.9: 77721512-77721512 on Build GRCh38
rs190985140 Chr.9: 77721576-77721576 on Build GRCh38
rs574519710 Chr.9: 77721565-77721565 on Build GRCh38
rs758215116 Chr.9: 77721422-77721422 on Build GRCh38
rs576775511 Chr.9: 77721572-77721572 on Build GRCh38
rs769315863 Chr.9: 77721486-77721486 on Build GRCh38
rs199754984 Chr.9: 77721464-77721464 on Build GRCh38
rs777546823 Chr.9: 77721492-77721492 on Build GRCh38
rs777234154 Chr.9: 77721550-77721550 on Build GRCh38
rs397825484 Chr.9: 77721439-77721439 on Build GRCh38
rs764430308 Chr.9: 77721413-77721413 on Build GRCh38
rs772749242 Chr.9: 77721499-77721499 on Build GRCh38
rs762604216 Chr.9: 77721518-77721518 on Build GRCh38
rs185734369 Chr.9: 77721551-77721551 on Build GRCh38
rs778106531 Chr.9: 77721533-77721534 on Build GRCh38
rs558410927 Chr.9: 77721526-77721526 on Build GRCh38
rs372746608 Chr.9: 77721469-77721469 on Build GRCh38
rs760152072 Chr.9: 77721497-77721497 on Build GRCh38
rs1059525 Chr.9: 77721416-77721416 on Build GRCh38
rs751391950 Chr.9: 77721533-77721533 on Build GRCh38
rs545662450 Chr.9: 77721540-77721541 on Build GRCh38
rs752142063 Chr.9: 77721547-77721547 on Build GRCh38

View Full Product Details