Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 116065456-116065729 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GFRA1
Entrez Gene ID
2674
Gene Name
GDNF family receptor alpha 1
Gene Aliases
GDNFR, GDNFRA, GFR-ALPHA-1, RET1L, RETL1, TRNR1
UniGene
Hs.388347
Gene Chromosome Location
Chr.10: 116056925-116273645 on Build GRCh38
Transcripts
NM_001145453.2 NM_005264.5 NM_145793.3 XM_005269696.1 XM_011539634.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1675591 c.1225T>G p.C409G Chr.10: 116065584-116065584 on Build GRCh38
4451459 c.1213A>T p.N405Y Chr.10: 116065596-116065596 on Build GRCh38
4451458 c.1228A>T p.N410Y Chr.10: 116065596-116065596 on Build GRCh38
6860463 c.1217C>G p.T406R Chr.10: 116065592-116065592 on Build GRCh38
6860464 c.869C>G p.T290R Chr.10: 116065592-116065592 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374656900 Chr.10: 116065600-116065600 on Build GRCh38
rs768946167 Chr.10: 116065524-116065524 on Build GRCh38
rs759451878 Chr.10: 116065547-116065547 on Build GRCh38
rs376337481 Chr.10: 116065546-116065546 on Build GRCh38
rs973732329 Chr.10: 116065540-116065540 on Build GRCh38
rs79252560 Chr.10: 116065618-116065618 on Build GRCh38
rs113238389 Chr.10: 116065659-116065659 on Build GRCh38
rs749335259 Chr.10: 116065601-116065601 on Build GRCh38
rs767978766 Chr.10: 116065642-116065642 on Build GRCh38
rs944178542 Chr.10: 116065627-116065627 on Build GRCh38
rs770814507 Chr.10: 116065656-116065656 on Build GRCh38
rs752134873 Chr.10: 116065554-116065554 on Build GRCh38
rs781398878 Chr.10: 116065581-116065581 on Build GRCh38
rs534173587 Chr.10: 116065526-116065526 on Build GRCh38
rs1024694145 Chr.10: 116065653-116065654 on Build GRCh38
rs897681332 Chr.10: 116065636-116065636 on Build GRCh38
rs746444341 Chr.10: 116065588-116065588 on Build GRCh38
rs375592850 Chr.10: 116065685-116065685 on Build GRCh38
rs758060117 Chr.10: 116065560-116065560 on Build GRCh38
rs769034000 Chr.10: 116065607-116065607 on Build GRCh38
rs371781884 Chr.10: 116065665-116065665 on Build GRCh38
rs371052783 Chr.10: 116065565-116065565 on Build GRCh38
rs750267988 Chr.10: 116065695-116065695 on Build GRCh38
rs775001184 Chr.10: 116065670-116065670 on Build GRCh38
rs1032464839 Chr.10: 116065693-116065693 on Build GRCh38
rs765100999 Chr.10: 116065668-116065668 on Build GRCh38
rs912731005 Chr.10: 116065612-116065612 on Build GRCh38
rs570225661 Chr.10: 116065641-116065642 on Build GRCh38
rs764791174 Chr.10: 116065548-116065548 on Build GRCh38
rs1036272843 Chr.10: 116065633-116065633 on Build GRCh38
rs1034658710 Chr.10: 116065487-116065487 on Build GRCh38
rs879095690 Chr.10: 116065509-116065509 on Build GRCh38
rs748293548 Chr.10: 116065655-116065655 on Build GRCh38
rs774676790 Chr.10: 116065616-116065616 on Build GRCh38
rs76253448 Chr.10: 116065595-116065595 on Build GRCh38
rs774589269 Chr.10: 116065565-116065566 on Build GRCh38
rs921543019 Chr.10: 116065700-116065700 on Build GRCh38
rs189945277 Chr.10: 116065578-116065578 on Build GRCh38
rs751898797 Chr.10: 116065572-116065572 on Build GRCh38
rs760482344 Chr.10: 116065529-116065529 on Build GRCh38

View Full Product Details