Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 121677150-121677423 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GOLGB1
Entrez Gene ID
2804
Gene Name
golgin B1
Gene Aliases
GCP, GCP372, GOLIM1
UniGene
Hs.213389
Gene Chromosome Location
Chr.3: 121663199-121749908 on Build GRCh38
Transcripts
NM_001256486.1 NM_001256487.1 NM_001256488.1 NM_004487.4 XM_005247371.4 XM_005247372.4 XM_005247373.1 XM_006713587.1 XM_006713588.1 XM_006713589.1 XM_006713590.1 XM_006713591.1 XM_011512699.2 XM_017006189.1 XM_017006190.1 XM_017006191.1 XM_017006192.1 XM_017006193.1 XM_017006194.1 XM_017006195.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6096033 c.8995C>T p.Q2999* Chr.3: 121677314-121677314 on Build GRCh38
7171620 c.9003C>G p.L3001L Chr.3: 121677306-121677306 on Build GRCh38
1418175 c.8957A>C p.N2986T Chr.3: 121677352-121677352 on Build GRCh38
1208464 c.8924C>T p.S2975L Chr.3: 121677385-121677385 on Build GRCh38
5594636 c.8962A>G p.I2988V Chr.3: 121677347-121677347 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148944271 Chr.3: 121677297-121677297 on Build GRCh38
rs762579926 Chr.3: 121677342-121677342 on Build GRCh38
rs751024357 Chr.3: 121677374-121677374 on Build GRCh38
rs943410058 Chr.3: 121677369-121677369 on Build GRCh38
rs777118617 Chr.3: 121677317-121677317 on Build GRCh38
rs745648998 Chr.3: 121677256-121677256 on Build GRCh38
rs919418303 Chr.3: 121677234-121677234 on Build GRCh38
rs774104640 Chr.3: 121677202-121677203 on Build GRCh38
rs372392359 Chr.3: 121677215-121677216 on Build GRCh38
rs78322216 Chr.3: 121677213-121677213 on Build GRCh38
rs752345956 Chr.3: 121677322-121677322 on Build GRCh38
rs773134594 Chr.3: 121677311-121677311 on Build GRCh38
rs757288764 Chr.3: 121677378-121677378 on Build GRCh38
rs1030384349 Chr.3: 121677218-121677218 on Build GRCh38
rs774500210 Chr.3: 121677270-121677270 on Build GRCh38
rs191079517 Chr.3: 121677263-121677263 on Build GRCh38
rs201451141 Chr.3: 121677252-121677252 on Build GRCh38
rs764992063 Chr.3: 121677319-121677319 on Build GRCh38
rs201378767 Chr.3: 121677213-121677213 on Build GRCh38
rs397841021 Chr.3: 121677212-121677213 on Build GRCh38
rs982039541 Chr.3: 121677215-121677215 on Build GRCh38
rs72587269 Chr.3: 121677211-121677212 on Build GRCh38
rs750859071 Chr.3: 121677361-121677372 on Build GRCh38
rs3835145 Chr.3: 121677203-121677204 on Build GRCh38
rs371475381 Chr.3: 121677318-121677318 on Build GRCh38
rs879016404 Chr.3: 121677198-121677198 on Build GRCh38
rs761292110 Chr.3: 121677316-121677316 on Build GRCh38
rs138361473 Chr.3: 121677325-121677327 on Build GRCh38
rs188723527 Chr.3: 121677222-121677222 on Build GRCh38
rs375764117 Chr.3: 121677347-121677347 on Build GRCh38
rs529915841 Chr.3: 121677287-121677287 on Build GRCh38

View Full Product Details