Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 125905109-125905376 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX25
Entrez Gene ID
29118
Gene Name
DEAD-box helicase 25
Gene Aliases
GRTH
UniGene
Hs.420263
Gene Chromosome Location
Chr.11: 125903275-125923110 on Build GRCh38
Transcripts
NM_013264.4 XM_011542790.2 XM_011542791.2 XM_017017626.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4522973 c.118G>T p.V40F Chr.11: 125905266-125905266 on Build GRCh38
924587 c.122G>A p.R41Q Chr.11: 125905270-125905270 on Build GRCh38
1352394 c.70A>G p.N24D Chr.11: 125905218-125905218 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs758524145 Chr.11: 125905268-125905268 on Build GRCh38
rs553193468 Chr.11: 125905276-125905276 on Build GRCh38
rs925677859 Chr.11: 125905348-125905348 on Build GRCh38
rs779956961 Chr.11: 125905270-125905270 on Build GRCh38
rs1029344487 Chr.11: 125905181-125905181 on Build GRCh38
rs760102486 Chr.11: 125905197-125905198 on Build GRCh38
rs1050142199 Chr.11: 125905330-125905330 on Build GRCh38
rs772513703 Chr.11: 125905201-125905202 on Build GRCh38
rs367864629 Chr.11: 125905281-125905281 on Build GRCh38
rs927401021 Chr.11: 125905210-125905210 on Build GRCh38
rs920405231 Chr.11: 125905301-125905301 on Build GRCh38
rs140716615 Chr.11: 125905136-125905136 on Build GRCh38
rs992851814 Chr.11: 125905249-125905249 on Build GRCh38
rs777894799 Chr.11: 125905204-125905204 on Build GRCh38
rs960286700 Chr.11: 125905221-125905221 on Build GRCh38
rs761871174 Chr.11: 125905280-125905280 on Build GRCh38
rs890219274 Chr.11: 125905333-125905333 on Build GRCh38
rs115679290 Chr.11: 125905344-125905344 on Build GRCh38
rs1009157248 Chr.11: 125905247-125905247 on Build GRCh38
rs954611135 Chr.11: 125905208-125905208 on Build GRCh38
rs996164745 Chr.11: 125905163-125905163 on Build GRCh38
rs931819033 Chr.11: 125905309-125905309 on Build GRCh38
rs899201784 Chr.11: 125905135-125905135 on Build GRCh38
rs907746898 Chr.11: 125905256-125905256 on Build GRCh38
rs1020166167 Chr.11: 125905259-125905259 on Build GRCh38
rs536712689 Chr.11: 125905261-125905261 on Build GRCh38
rs548065485 Chr.11: 125905355-125905355 on Build GRCh38
rs978529369 Chr.11: 125905345-125905345 on Build GRCh38

View Full Product Details