Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 4359321-4359548 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CORO7
Entrez Gene ID
79585
Gene Name
coronin 7
Gene Aliases
0610011B16Rik, CRN7, POD1
UniGene
Hs.437957
Gene Chromosome Location
Chr.16: 4354542-4416961 on Build GRCh38
Transcripts
NM_001201472.1 NM_001201473.1 NM_024535.4 NM_001201479.1

Gene Symbol
CORO7-PAM16
Entrez Gene ID
100529144
Gene Name
CORO7-PAM16 readthrough
Gene Aliases
-
UniGene
Hs.437957
Gene Chromosome Location
Chr.16: 4340251-4416961 on Build GRCh38
Transcripts
NM_001201472.1 NM_001201473.1 NM_024535.4 NM_001201479.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3402326 c.2261G>A p.R754H Chr.16: 4359375-4359375 on Build GRCh38
6197066 c.2260C>T p.R754C Chr.16: 4359376-4359376 on Build GRCh38
4847567 c.2215G>C p.D739H Chr.16: 4359515-4359515 on Build GRCh38
6634290 c.2215G>A p.D739N Chr.16: 4359515-4359515 on Build GRCh38
6866687 c.2224A>T p.T742S Chr.16: 4359506-4359506 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs529198905 Chr.16: 4359390-4359390 on Build GRCh38
rs376945155 Chr.16: 4359500-4359500 on Build GRCh38
rs569923937 Chr.16: 4359440-4359440 on Build GRCh38
rs771101075 Chr.16: 4359505-4359505 on Build GRCh38
rs746971981 Chr.16: 4359415-4359415 on Build GRCh38
rs778330329 Chr.16: 4359358-4359358 on Build GRCh38
rs905412530 Chr.16: 4359463-4359463 on Build GRCh38
rs184824335 Chr.16: 4359425-4359425 on Build GRCh38
rs752123001 Chr.16: 4359387-4359387 on Build GRCh38
rs367809869 Chr.16: 4359367-4359367 on Build GRCh38
rs770075857 Chr.16: 4359433-4359433 on Build GRCh38
rs760398003 Chr.16: 4359366-4359366 on Build GRCh38
rs770430395 Chr.16: 4359357-4359357 on Build GRCh38
rs375902989 Chr.16: 4359361-4359361 on Build GRCh38
rs200938494 Chr.16: 4359523-4359533 on Build GRCh38
rs762215165 Chr.16: 4359432-4359433 on Build GRCh38
rs746919528 Chr.16: 4359485-4359485 on Build GRCh38
rs142159766 Chr.16: 4359515-4359515 on Build GRCh38
rs781034417 Chr.16: 4359490-4359490 on Build GRCh38
rs771575935 Chr.16: 4359412-4359412 on Build GRCh38
rs755396280 Chr.16: 4359349-4359349 on Build GRCh38
rs373505885 Chr.16: 4359477-4359477 on Build GRCh38
rs757855791 Chr.16: 4359481-4359481 on Build GRCh38
rs886157280 Chr.16: 4359466-4359466 on Build GRCh38
rs772878441 Chr.16: 4359469-4359485 on Build GRCh38
rs763066448 Chr.16: 4359433-4359433 on Build GRCh38
rs764477388 Chr.16: 4359461-4359461 on Build GRCh38
rs776338024 Chr.16: 4359417-4359417 on Build GRCh38
rs775263625 Chr.16: 4359365-4359365 on Build GRCh38
rs971451141 Chr.16: 4359399-4359399 on Build GRCh38
rs547483612 Chr.16: 4359383-4359383 on Build GRCh38
rs144121826 Chr.16: 4359438-4359438 on Build GRCh38
rs113640236 Chr.16: 4359512-4359512 on Build GRCh38
rs140974533 Chr.16: 4359350-4359350 on Build GRCh38
rs761973189 Chr.16: 4359370-4359370 on Build GRCh38
rs376933725 Chr.16: 4359376-4359376 on Build GRCh38
rs777316531 Chr.16: 4359433-4359442 on Build GRCh38
rs201343195 Chr.16: 4359362-4359362 on Build GRCh38
rs377174017 Chr.16: 4359418-4359418 on Build GRCh38
rs755343378 Chr.16: 4359389-4359389 on Build GRCh38
rs778475649 Chr.16: 4359396-4359396 on Build GRCh38
rs562307467 Chr.16: 4359412-4359412 on Build GRCh38
rs372039186 Chr.16: 4359368-4359368 on Build GRCh38
rs747462056 Chr.16: 4359342-4359350 on Build GRCh38
rs369568897 Chr.16: 4359416-4359416 on Build GRCh38
rs766586397 Chr.16: 4359436-4359436 on Build GRCh38
rs931932531 Chr.16: 4359363-4359363 on Build GRCh38
rs750090306 Chr.16: 4359460-4359460 on Build GRCh38
rs756913162 Chr.16: 4359395-4359395 on Build GRCh38
rs188003154 Chr.16: 4359516-4359516 on Build GRCh38
rs150350387 Chr.16: 4359501-4359501 on Build GRCh38
rs745439199 Chr.16: 4359404-4359404 on Build GRCh38
rs765337019 Chr.16: 4359375-4359375 on Build GRCh38
rs764733122 Chr.16: 4359458-4359458 on Build GRCh38
rs1046586414 Chr.16: 4359439-4359439 on Build GRCh38
rs538517913 Chr.16: 4359486-4359486 on Build GRCh38
rs1048361218 Chr.16: 4359499-4359499 on Build GRCh38
rs199853197 Chr.16: 4359345-4359345 on Build GRCh38
rs536234389 Chr.16: 4359426-4359426 on Build GRCh38
rs779910359 Chr.16: 4359414-4359414 on Build GRCh38
rs779572564 Chr.16: 4359482-4359482 on Build GRCh38

View Full Product Details