Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 3444098-3444349 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HGFAC
Entrez Gene ID
3083
Gene Name
HGF activator
Gene Aliases
HGFA
UniGene
Hs.104
Gene Chromosome Location
Chr.4: 3441933-3449495 on Build GRCh38
Transcripts
NM_001297439.1 NM_001528.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6321114 c.625G>T p.E209* Chr.4: 3444337-3444337 on Build GRCh38
447802 c.591C>T p.C197C Chr.4: 3444154-3444154 on Build GRCh38
6039484 c.636delG p.G214fs*33 Chr.4: 3444348-3444348 on Build GRCh38
5401117 c.570G>A p.R190R Chr.4: 3444133-3444133 on Build GRCh38
4849505 c.613G>T p.E205* Chr.4: 3444325-3444325 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759716394 Chr.4: 3444144-3444144 on Build GRCh38
rs375515577 Chr.4: 3444184-3444184 on Build GRCh38
rs1028905187 Chr.4: 3444204-3444204 on Build GRCh38
rs776565154 Chr.4: 3444142-3444142 on Build GRCh38
rs890417453 Chr.4: 3444211-3444211 on Build GRCh38
rs367852767 Chr.4: 3444149-3444149 on Build GRCh38
rs763286183 Chr.4: 3444191-3444191 on Build GRCh38
rs763372648 Chr.4: 3444150-3444150 on Build GRCh38
rs761304367 Chr.4: 3444179-3444181 on Build GRCh38
rs867644897 Chr.4: 3444213-3444213 on Build GRCh38
rs373167210 Chr.4: 3444154-3444154 on Build GRCh38
rs535094909 Chr.4: 3444164-3444164 on Build GRCh38
rs754395468 Chr.4: 3444264-3444264 on Build GRCh38
rs768037340 Chr.4: 3444174-3444175 on Build GRCh38
rs754566283 Chr.4: 3444171-3444171 on Build GRCh38
rs539282936 Chr.4: 3444134-3444134 on Build GRCh38
rs370355597 Chr.4: 3444130-3444130 on Build GRCh38
rs571948667 Chr.4: 3444239-3444239 on Build GRCh38
rs188255678 Chr.4: 3444256-3444256 on Build GRCh38
rs199726049 Chr.4: 3444155-3444155 on Build GRCh38
rs769950010 Chr.4: 3444188-3444188 on Build GRCh38
rs747818555 Chr.4: 3444174-3444174 on Build GRCh38
rs923793961 Chr.4: 3444167-3444167 on Build GRCh38
rs368328359 Chr.4: 3444189-3444189 on Build GRCh38
rs746868651 Chr.4: 3444176-3444176 on Build GRCh38
rs752279697 Chr.4: 3444283-3444283 on Build GRCh38
rs777455883 Chr.4: 3444304-3444304 on Build GRCh38
rs764491214 Chr.4: 3444200-3444200 on Build GRCh38
rs989384328 Chr.4: 3444198-3444198 on Build GRCh38
rs906669169 Chr.4: 3444240-3444240 on Build GRCh38
rs898958951 Chr.4: 3444183-3444183 on Build GRCh38
rs758130108 Chr.4: 3444175-3444175 on Build GRCh38
rs757848185 Chr.4: 3444160-3444160 on Build GRCh38
rs776260967 Chr.4: 3444167-3444168 on Build GRCh38
rs550921879 Chr.4: 3444305-3444305 on Build GRCh38
rs200391858 Chr.4: 3444263-3444263 on Build GRCh38
rs377317114 Chr.4: 3444331-3444331 on Build GRCh38
rs776139494 Chr.4: 3444177-3444178 on Build GRCh38
rs140416920 Chr.4: 3444131-3444131 on Build GRCh38
rs778686460 Chr.4: 3444129-3444129 on Build GRCh38
rs761239456 Chr.4: 3444168-3444169 on Build GRCh38
rs768044501 Chr.4: 3444166-3444167 on Build GRCh38
rs745902639 Chr.4: 3444186-3444186 on Build GRCh38
rs770756848 Chr.4: 3444180-3444180 on Build GRCh38
rs1042531466 Chr.4: 3444214-3444214 on Build GRCh38
rs753274559 Chr.4: 3444169-3444169 on Build GRCh38
rs570278133 Chr.4: 3444132-3444132 on Build GRCh38
rs150376439 Chr.4: 3444137-3444137 on Build GRCh38
rs372484613 Chr.4: 3444298-3444298 on Build GRCh38
rs898027025 Chr.4: 3444278-3444278 on Build GRCh38
rs201128686 Chr.4: 3444148-3444148 on Build GRCh38
rs764524869 Chr.4: 3444189-3444190 on Build GRCh38
rs555301746 Chr.4: 3444201-3444201 on Build GRCh38
rs35018889 Chr.4: 3444330-3444330 on Build GRCh38
rs532318836 Chr.4: 3444257-3444257 on Build GRCh38
rs746988281 Chr.4: 3444139-3444139 on Build GRCh38
rs763028128 Chr.4: 3444308-3444309 on Build GRCh38
rs769417510 Chr.4: 3444170-3444171 on Build GRCh38
rs759388518 Chr.4: 3444244-3444244 on Build GRCh38
rs867458853 Chr.4: 3444120-3444120 on Build GRCh38
rs373809126 Chr.4: 3444147-3444147 on Build GRCh38
rs762297146 Chr.4: 3444203-3444203 on Build GRCh38
rs767890763 Chr.4: 3444212-3444212 on Build GRCh38
rs917199559 Chr.4: 3444143-3444143 on Build GRCh38
rs773025649 Chr.4: 3444171-3444181 on Build GRCh38
rs764815435 Chr.4: 3444269-3444269 on Build GRCh38

View Full Product Details