Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 131274906-131275135 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGSF1
Entrez Gene ID
3547
Gene Name
immunoglobulin superfamily member 1
Gene Aliases
CHTE, IGCD1, IGDC1, INHBP, PGSF2, p120
UniGene
Hs.22111
Gene Chromosome Location
Chr.X: 131273506-131289457 on Build GRCh38
Transcripts
NM_001170961.1 NM_001170962.1 NM_001555.4 XM_011531330.1 XM_011531333.1 XM_011531334.1

SNP Detail

SNP ID Chromosome Location
rs776163735 Chr.X: 131274968-131274968 on Build GRCh38
rs777067808 Chr.X: 131275015-131275015 on Build GRCh38
rs143211470 Chr.X: 131275095-131275095 on Build GRCh38
rs371861555 Chr.X: 131274949-131274949 on Build GRCh38
rs762232691 Chr.X: 131275067-131275067 on Build GRCh38
rs956426611 Chr.X: 131274936-131274936 on Build GRCh38
rs267606354 Chr.X: 131275029-131275029 on Build GRCh38
rs745459312 Chr.X: 131274970-131274970 on Build GRCh38
rs986271107 Chr.X: 131275000-131275000 on Build GRCh38
rs774949449 Chr.X: 131275012-131275012 on Build GRCh38
rs376770776 Chr.X: 131274967-131274967 on Build GRCh38
rs745330779 Chr.X: 131274977-131274977 on Build GRCh38
rs190720568 Chr.X: 131274961-131274961 on Build GRCh38
rs750746517 Chr.X: 131275055-131275055 on Build GRCh38
rs988195384 Chr.X: 131274938-131274938 on Build GRCh38
rs909743754 Chr.X: 131275094-131275094 on Build GRCh38
rs764728624 Chr.X: 131275003-131275003 on Build GRCh38
rs371811870 Chr.X: 131274989-131274989 on Build GRCh38
rs369776380 Chr.X: 131274975-131274975 on Build GRCh38
rs767853834 Chr.X: 131275052-131275052 on Build GRCh38
rs753954641 Chr.X: 131275104-131275104 on Build GRCh38
rs764754381 Chr.X: 131275075-131275075 on Build GRCh38
rs762097558 Chr.X: 131275037-131275037 on Build GRCh38
rs775198831 Chr.X: 131274978-131274978 on Build GRCh38

View Full Product Details