Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 212068528-212068753 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DTL
Entrez Gene ID
51514
Gene Name
denticleless E3 ubiquitin protein ligase homolog
Gene Aliases
CDT2, DCAF2, L2DTL, RAMP
UniGene
Hs.656473
Gene Chromosome Location
Chr.1: 212035553-212105006 on Build GRCh38
Transcripts
NM_001286229.1 NM_001286230.1 NM_016448.3 XM_011509614.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1748140 c.912G>A p.K304K Chr.1: 212068693-212068693 on Build GRCh38
6841039 c.869G>A p.C290Y Chr.1: 212068650-212068650 on Build GRCh38
1748139 c.909G>A p.L303L Chr.1: 212068690-212068690 on Build GRCh38
5907878 c.818-4C>T p.? Chr.1: 212068595-212068595 on Build GRCh38
903538 c.856T>G p.L286V Chr.1: 212068637-212068637 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780305649 Chr.1: 212068673-212068673 on Build GRCh38
rs773081177 Chr.1: 212068579-212068579 on Build GRCh38
rs770376180 Chr.1: 212068727-212068727 on Build GRCh38
rs757624199 Chr.1: 212068635-212068635 on Build GRCh38
rs74378223 Chr.1: 212068606-212068606 on Build GRCh38
rs746948573 Chr.1: 212068721-212068721 on Build GRCh38
rs762716704 Chr.1: 212068730-212068730 on Build GRCh38
rs368819236 Chr.1: 212068580-212068580 on Build GRCh38
rs771520378 Chr.1: 212068572-212068572 on Build GRCh38
rs373190277 Chr.1: 212068570-212068570 on Build GRCh38
rs528954322 Chr.1: 212068718-212068718 on Build GRCh38
rs780395579 Chr.1: 212068566-212068566 on Build GRCh38
rs755257350 Chr.1: 212068680-212068680 on Build GRCh38
rs925284222 Chr.1: 212068670-212068670 on Build GRCh38
rs34452090 Chr.1: 212068657-212068657 on Build GRCh38
rs978191417 Chr.1: 212068715-212068715 on Build GRCh38
rs759206508 Chr.1: 212068625-212068625 on Build GRCh38
rs751952843 Chr.1: 212068632-212068632 on Build GRCh38
rs373514199 Chr.1: 212068587-212068587 on Build GRCh38
rs776222903 Chr.1: 212068614-212068614 on Build GRCh38
rs990905702 Chr.1: 212068712-212068712 on Build GRCh38
rs781442911 Chr.1: 212068731-212068732 on Build GRCh38
rs770074439 Chr.1: 212068627-212068627 on Build GRCh38
rs771809491 Chr.1: 212068709-212068709 on Build GRCh38
rs375718174 Chr.1: 212068571-212068571 on Build GRCh38
rs745951601 Chr.1: 212068563-212068563 on Build GRCh38
rs139513421 Chr.1: 212068700-212068700 on Build GRCh38
rs561986101 Chr.1: 212068674-212068674 on Build GRCh38
rs375048503 Chr.1: 212068720-212068720 on Build GRCh38
rs750936151 Chr.1: 212068658-212068658 on Build GRCh38
rs779070233 Chr.1: 212068694-212068694 on Build GRCh38
rs764380420 Chr.1: 212068629-212068629 on Build GRCh38
rs755173703 Chr.1: 212068603-212068604 on Build GRCh38
rs756223929 Chr.1: 212068663-212068663 on Build GRCh38
rs201883312 Chr.1: 212068565-212068565 on Build GRCh38

View Full Product Details