Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 100535908-100536181 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIF1AN
Entrez Gene ID
55662
Gene Name
hypoxia inducible factor 1 alpha subunit inhibitor
Gene Aliases
FIH1
UniGene
Hs.500788
Gene Chromosome Location
Chr.10: 100535884-100553924 on Build GRCh38
Transcripts
NM_017902.2 XM_011539940.2 XM_011539941.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6260439 c.177+3G>T p.? Chr.10: 100536138-100536138 on Build GRCh38
3741461 c.112C>A p.P38T Chr.10: 100536070-100536070 on Build GRCh38
4787532 c.27G>A p.V9V Chr.10: 100535985-100535985 on Build GRCh38
426936 c.121C>G p.P41A Chr.10: 100536079-100536079 on Build GRCh38
6248107 c.131G>T p.R44L Chr.10: 100536089-100536089 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774739059 Chr.10: 100536110-100536110 on Build GRCh38
rs368006181 Chr.10: 100535935-100535935 on Build GRCh38
rs768819336 Chr.10: 100536107-100536107 on Build GRCh38
rs775707186 Chr.10: 100535993-100535993 on Build GRCh38
rs915401255 Chr.10: 100536105-100536105 on Build GRCh38
rs749421411 Chr.10: 100536080-100536080 on Build GRCh38
rs750905299 Chr.10: 100535946-100535946 on Build GRCh38
rs769829095 Chr.10: 100536069-100536069 on Build GRCh38
rs768180437 Chr.10: 100535942-100535942 on Build GRCh38
rs1035016923 Chr.10: 100535988-100535988 on Build GRCh38
rs745645333 Chr.10: 100536063-100536063 on Build GRCh38
rs760930335 Chr.10: 100536129-100536129 on Build GRCh38
rs1009234647 Chr.10: 100535947-100535947 on Build GRCh38
rs758109470 Chr.10: 100535963-100535963 on Build GRCh38
rs746852281 Chr.10: 100535968-100535968 on Build GRCh38
rs757582822 Chr.10: 100536102-100536102 on Build GRCh38
rs752259488 Chr.10: 100536047-100536047 on Build GRCh38
rs906211591 Chr.10: 100535975-100535975 on Build GRCh38
rs762302836 Chr.10: 100536009-100536009 on Build GRCh38
rs762073529 Chr.10: 100536118-100536118 on Build GRCh38
rs758958755 Chr.10: 100535949-100535950 on Build GRCh38
rs764407611 Chr.10: 100535928-100535928 on Build GRCh38
rs370894479 Chr.10: 100536050-100536050 on Build GRCh38
rs774685992 Chr.10: 100536007-100536007 on Build GRCh38
rs761173401 Chr.10: 100536024-100536024 on Build GRCh38
rs753212884 Chr.10: 100535947-100535948 on Build GRCh38
rs766955833 Chr.10: 100536146-100536146 on Build GRCh38
rs776463635 Chr.10: 100535977-100535977 on Build GRCh38
rs745770096 Chr.10: 100535980-100535980 on Build GRCh38
rs781218525 Chr.10: 100536138-100536138 on Build GRCh38
rs989706394 Chr.10: 100536102-100536102 on Build GRCh38
rs892244975 Chr.10: 100535942-100535944 on Build GRCh38
rs778432940 Chr.10: 100535953-100535953 on Build GRCh38
rs899642316 Chr.10: 100535927-100535927 on Build GRCh38
rs754310456 Chr.10: 100536156-100536156 on Build GRCh38
rs770680594 Chr.10: 100535973-100535973 on Build GRCh38
rs377650841 Chr.10: 100536031-100536031 on Build GRCh38
rs780983095 Chr.10: 100536062-100536062 on Build GRCh38
rs778010961 Chr.10: 100535961-100535963 on Build GRCh38
rs140171051 Chr.10: 100536051-100536051 on Build GRCh38
rs751171143 Chr.10: 100536057-100536057 on Build GRCh38
rs777387138 Chr.10: 100535966-100535966 on Build GRCh38
rs754472955 Chr.10: 100535951-100535951 on Build GRCh38
rs747755694 Chr.10: 100535954-100535954 on Build GRCh38
rs769768445 Chr.10: 100535987-100535987 on Build GRCh38
rs757065092 Chr.10: 100536060-100536060 on Build GRCh38
rs2295778 Chr.10: 100536079-100536079 on Build GRCh38
rs763113624 Chr.10: 100535994-100535994 on Build GRCh38
rs1016507563 Chr.10: 100535997-100535997 on Build GRCh38
rs373246853 Chr.10: 100536015-100536015 on Build GRCh38
rs974639762 Chr.10: 100536022-100536022 on Build GRCh38
rs764462537 Chr.10: 100536004-100536004 on Build GRCh38
rs1024675396 Chr.10: 100535972-100535972 on Build GRCh38
rs773689188 Chr.10: 100536128-100536128 on Build GRCh38
rs772332480 Chr.10: 100536124-100536124 on Build GRCh38
rs750911950 Chr.10: 100536023-100536023 on Build GRCh38
rs747018581 Chr.10: 100536009-100536011 on Build GRCh38

View Full Product Details