Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 67441773-67442014 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CORO1B
Entrez Gene ID
57175
Gene Name
coronin 1B
Gene Aliases
CORONIN-2
UniGene
Hs.6191
Gene Chromosome Location
Chr.11: 67438047-67443821 on Build GRCh38
Transcripts
NM_001018070.2 NM_020441.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6634206 c.295G>T p.A99S Chr.11: 67441995-67441995 on Build GRCh38
7362405 c.367G>C p.E123Q Chr.11: 67441820-67441820 on Build GRCh38
7362404 c.367G>C p.E123Q Chr.11: 67441820-67441820 on Build GRCh38
7237708 c.352A>C p.T118P Chr.11: 67441835-67441835 on Build GRCh38
7237706 c.352A>C p.T118P Chr.11: 67441835-67441835 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780259406 Chr.11: 67441986-67441988 on Build GRCh38
rs933105006 Chr.11: 67441963-67441963 on Build GRCh38
rs373313261 Chr.11: 67441891-67441891 on Build GRCh38
rs371393269 Chr.11: 67441952-67441952 on Build GRCh38
rs779400132 Chr.11: 67441860-67441860 on Build GRCh38
rs753834156 Chr.11: 67441909-67441909 on Build GRCh38
rs145707942 Chr.11: 67441820-67441820 on Build GRCh38
rs754173098 Chr.11: 67441987-67441988 on Build GRCh38
rs759263540 Chr.11: 67441816-67441816 on Build GRCh38
rs200382627 Chr.11: 67441935-67441935 on Build GRCh38
rs758707123 Chr.11: 67441973-67441973 on Build GRCh38
rs752039855 Chr.11: 67441984-67441984 on Build GRCh38
rs759802813 Chr.11: 67441950-67441950 on Build GRCh38
rs778279504 Chr.11: 67441977-67441977 on Build GRCh38
rs143397996 Chr.11: 67441923-67441923 on Build GRCh38
rs559490630 Chr.11: 67441884-67441884 on Build GRCh38
rs372470733 Chr.11: 67441815-67441815 on Build GRCh38
rs1017917152 Chr.11: 67441835-67441835 on Build GRCh38
rs774168938 Chr.11: 67441939-67441939 on Build GRCh38
rs756543084 Chr.11: 67441954-67441955 on Build GRCh38
rs760740853 Chr.11: 67441942-67441942 on Build GRCh38
rs776228956 Chr.11: 67441895-67441895 on Build GRCh38
rs765622939 Chr.11: 67441959-67441959 on Build GRCh38
rs778205713 Chr.11: 67441885-67441885 on Build GRCh38
rs762852329 Chr.11: 67441828-67441828 on Build GRCh38
rs148916908 Chr.11: 67441854-67441854 on Build GRCh38
rs942935140 Chr.11: 67441985-67441985 on Build GRCh38
rs200415950 Chr.11: 67441989-67441989 on Build GRCh38
rs563841106 Chr.11: 67441943-67441943 on Build GRCh38
rs548608950 Chr.11: 67441849-67441849 on Build GRCh38
rs541464028 Chr.11: 67441842-67441842 on Build GRCh38
rs746572332 Chr.11: 67441800-67441800 on Build GRCh38
rs1046224269 Chr.11: 67441937-67441937 on Build GRCh38
rs770607931 Chr.11: 67441806-67441806 on Build GRCh38
rs956062890 Chr.11: 67441876-67441876 on Build GRCh38
rs768624407 Chr.11: 67441936-67441936 on Build GRCh38
rs754022069 Chr.11: 67441944-67441944 on Build GRCh38
rs780654908 Chr.11: 67441990-67441990 on Build GRCh38
rs763974640 Chr.11: 67441841-67441841 on Build GRCh38
rs754115790 Chr.11: 67441853-67441853 on Build GRCh38
rs753102227 Chr.11: 67441972-67441972 on Build GRCh38
rs375496989 Chr.11: 67441827-67441827 on Build GRCh38
rs770355835 Chr.11: 67441894-67441894 on Build GRCh38
rs369902133 Chr.11: 67441883-67441883 on Build GRCh38
rs769482342 Chr.11: 67441915-67441915 on Build GRCh38

View Full Product Details