Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 2794525-2794750 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPXM1
Entrez Gene ID
56265
Gene Name
carboxypeptidase X (M14 family), member 1
Gene Aliases
CPX1, CPXM
UniGene
Hs.659346
Gene Chromosome Location
Chr.20: 2794069-2800646 on Build GRCh38
Transcripts
NM_001184699.1 NM_019609.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6635005 c.1962G>A p.T654T Chr.20: 2794538-2794538 on Build GRCh38
6092945 c.1911G>A p.G637G Chr.20: 2794589-2794589 on Build GRCh38
1615425 c.1861-2A>T p.? Chr.20: 2794641-2794641 on Build GRCh38
477944 c.1935G>A p.V645V Chr.20: 2794565-2794565 on Build GRCh38
4532644 c.1902G>A p.T634T Chr.20: 2794598-2794598 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760210168 Chr.20: 2794636-2794636 on Build GRCh38
rs1053274396 Chr.20: 2794564-2794564 on Build GRCh38
rs971359526 Chr.20: 2794719-2794719 on Build GRCh38
rs749606246 Chr.20: 2794607-2794607 on Build GRCh38
rs762194038 Chr.20: 2794625-2794625 on Build GRCh38
rs768886563 Chr.20: 2794610-2794610 on Build GRCh38
rs774844112 Chr.20: 2794611-2794611 on Build GRCh38
rs772660245 Chr.20: 2794635-2794635 on Build GRCh38
rs780336421 Chr.20: 2794687-2794687 on Build GRCh38
rs778854153 Chr.20: 2794572-2794572 on Build GRCh38
rs267605867 Chr.20: 2794561-2794561 on Build GRCh38
rs369610590 Chr.20: 2794567-2794567 on Build GRCh38
rs753591181 Chr.20: 2794553-2794553 on Build GRCh38
rs538540341 Chr.20: 2794676-2794676 on Build GRCh38
rs142917655 Chr.20: 2794634-2794634 on Build GRCh38
rs758392407 Chr.20: 2794665-2794665 on Build GRCh38
rs1025682083 Chr.20: 2794671-2794671 on Build GRCh38
rs372906016 Chr.20: 2794546-2794546 on Build GRCh38
rs546064620 Chr.20: 2794721-2794721 on Build GRCh38
rs757362967 Chr.20: 2794675-2794675 on Build GRCh38
rs781393311 Chr.20: 2794584-2794584 on Build GRCh38
rs752581713 Chr.20: 2794664-2794664 on Build GRCh38
rs145964876 Chr.20: 2794598-2794598 on Build GRCh38
rs754719547 Chr.20: 2794651-2794651 on Build GRCh38
rs142423261 Chr.20: 2794594-2794594 on Build GRCh38
rs371978601 Chr.20: 2794648-2794648 on Build GRCh38
rs748025686 Chr.20: 2794579-2794579 on Build GRCh38
rs139729690 Chr.20: 2794627-2794627 on Build GRCh38
rs764939539 Chr.20: 2794659-2794659 on Build GRCh38
rs758432124 Chr.20: 2794580-2794580 on Build GRCh38
rs127078 Chr.20: 2794686-2794686 on Build GRCh38
rs960504973 Chr.20: 2794716-2794716 on Build GRCh38
rs113052252 Chr.20: 2794547-2794547 on Build GRCh38
rs765950367 Chr.20: 2794637-2794637 on Build GRCh38
rs1051222640 Chr.20: 2794554-2794554 on Build GRCh38
rs148259975 Chr.20: 2794590-2794590 on Build GRCh38
rs375368164 Chr.20: 2794557-2794557 on Build GRCh38

View Full Product Details