Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 154275109-154275342 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HAX1
Entrez Gene ID
10456
Gene Name
HCLS1 associated protein X-1
Gene Aliases
HCLSBP1, HS1BP1, SCN3
UniGene
Hs.199625
Gene Chromosome Location
Chr.1: 154272563-154275875 on Build GRCh38
Transcripts
NM_001018837.1 NM_006118.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6121542 c.646A>G p.I216V Chr.1: 154275243-154275243 on Build GRCh38
5843786 c.655C>T p.P219S Chr.1: 154275252-154275252 on Build GRCh38
5887575 c.566C>T p.S189F Chr.1: 154275163-154275163 on Build GRCh38
7370687 c.593C>A p.P198Q Chr.1: 154275190-154275190 on Build GRCh38
1600874 c.582G>A p.E194E Chr.1: 154275179-154275179 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776550825 Chr.1: 154275148-154275148 on Build GRCh38
rs1041678008 Chr.1: 154275202-154275202 on Build GRCh38
rs779277098 Chr.1: 154275255-154275255 on Build GRCh38
rs200327434 Chr.1: 154275174-154275174 on Build GRCh38
rs1024291441 Chr.1: 154275168-154275168 on Build GRCh38
rs757198202 Chr.1: 154275146-154275146 on Build GRCh38
rs746250096 Chr.1: 154275257-154275257 on Build GRCh38
rs768742603 Chr.1: 154275137-154275137 on Build GRCh38
rs199780741 Chr.1: 154275236-154275236 on Build GRCh38
rs748160893 Chr.1: 154275304-154275304 on Build GRCh38
rs575621654 Chr.1: 154275271-154275271 on Build GRCh38
rs146152769 Chr.1: 154275190-154275190 on Build GRCh38
rs372128634 Chr.1: 154275308-154275308 on Build GRCh38
rs777395523 Chr.1: 154275214-154275214 on Build GRCh38
rs760842693 Chr.1: 154275136-154275136 on Build GRCh38
rs776836844 Chr.1: 154275303-154275303 on Build GRCh38
rs146312637 Chr.1: 154275233-154275233 on Build GRCh38
rs755648319 Chr.1: 154275209-154275209 on Build GRCh38
rs74315322 Chr.1: 154275165-154275165 on Build GRCh38
rs999832713 Chr.1: 154275294-154275294 on Build GRCh38
rs1050683276 Chr.1: 154275135-154275135 on Build GRCh38
rs780455003 Chr.1: 154275266-154275266 on Build GRCh38
rs41313932 Chr.1: 154275261-154275261 on Build GRCh38
rs751426915 Chr.1: 154275171-154275171 on Build GRCh38
rs768728912 Chr.1: 154275300-154275300 on Build GRCh38
rs546841133 Chr.1: 154275162-154275162 on Build GRCh38
rs759265854 Chr.1: 154275183-154275183 on Build GRCh38
rs752302439 Chr.1: 154275191-154275191 on Build GRCh38
rs770912376 Chr.1: 154275305-154275305 on Build GRCh38
rs753259930 Chr.1: 154275223-154275223 on Build GRCh38

View Full Product Details