Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 69005272-69005531 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MRGPRF
Entrez Gene ID
116535
Gene Name
MAS related GPR family member F
Gene Aliases
GPR140, GPR168, MRGF, RTA
UniGene
Hs.118513
Gene Chromosome Location
Chr.11: 69004375-69013382 on Build GRCh38
Transcripts
NM_001098515.1 NM_145015.4 XM_017017170.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6500184 c.890A>C p.D297A Chr.11: 69005420-69005420 on Build GRCh38
5637184 c.903delG p.L302fs*>42 Chr.11: 69005407-69005407 on Build GRCh38
4509511 c.809C>T p.P270L Chr.11: 69005501-69005501 on Build GRCh38
6564837 c.798G>A p.Q266Q Chr.11: 69005512-69005512 on Build GRCh38
4509582 c.810C>T p.P270P Chr.11: 69005500-69005500 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760855839 Chr.11: 69005454-69005454 on Build GRCh38
rs1021362782 Chr.11: 69005417-69005417 on Build GRCh38
rs756421559 Chr.11: 69005361-69005361 on Build GRCh38
rs568789013 Chr.11: 69005411-69005411 on Build GRCh38
rs750813142 Chr.11: 69005360-69005360 on Build GRCh38
rs374094839 Chr.11: 69005503-69005503 on Build GRCh38
rs776664188 Chr.11: 69005341-69005341 on Build GRCh38
rs558204229 Chr.11: 69005294-69005294 on Build GRCh38
rs199870549 Chr.11: 69005403-69005403 on Build GRCh38
rs775241086 Chr.11: 69005497-69005497 on Build GRCh38
rs371144082 Chr.11: 69005362-69005362 on Build GRCh38
rs200224198 Chr.11: 69005397-69005397 on Build GRCh38
rs747885460 Chr.11: 69005311-69005311 on Build GRCh38
rs750602590 Chr.11: 69005442-69005442 on Build GRCh38
rs755456887 Chr.11: 69005366-69005366 on Build GRCh38
rs866609582 Chr.11: 69005302-69005302 on Build GRCh38
rs776539629 Chr.11: 69005406-69005406 on Build GRCh38
rs773126683 Chr.11: 69005329-69005329 on Build GRCh38
rs759601454 Chr.11: 69005409-69005409 on Build GRCh38
rs1045135378 Chr.11: 69005327-69005327 on Build GRCh38
rs374158773 Chr.11: 69005473-69005473 on Build GRCh38
rs866706538 Chr.11: 69005340-69005340 on Build GRCh38
rs61738965 Chr.11: 69005428-69005428 on Build GRCh38
rs377740520 Chr.11: 69005359-69005359 on Build GRCh38
rs184076521 Chr.11: 69005487-69005487 on Build GRCh38
rs777236371 Chr.11: 69005388-69005388 on Build GRCh38
rs778545345 Chr.11: 69005368-69005368 on Build GRCh38
rs756613938 Chr.11: 69005290-69005290 on Build GRCh38
rs552291439 Chr.11: 69005348-69005348 on Build GRCh38
rs1031429929 Chr.11: 69005444-69005444 on Build GRCh38
rs979475525 Chr.11: 69005443-69005443 on Build GRCh38
rs767801886 Chr.11: 69005438-69005438 on Build GRCh38
rs760483076 Chr.11: 69005330-69005330 on Build GRCh38
rs370955780 Chr.11: 69005490-69005490 on Build GRCh38
rs766777260 Chr.11: 69005459-69005459 on Build GRCh38
rs1007663493 Chr.11: 69005413-69005413 on Build GRCh38
rs564711813 Chr.11: 69005386-69005386 on Build GRCh38
rs752267382 Chr.11: 69005483-69005483 on Build GRCh38
rs746826672 Chr.11: 69005395-69005395 on Build GRCh38
rs769780377 Chr.11: 69005494-69005494 on Build GRCh38
rs771610988 Chr.11: 69005312-69005312 on Build GRCh38
rs747607052 Chr.11: 69005376-69005376 on Build GRCh38
rs754189845 Chr.11: 69005460-69005460 on Build GRCh38
rs771009282 Chr.11: 69005332-69005332 on Build GRCh38
rs758658446 Chr.11: 69005427-69005427 on Build GRCh38
rs144312357 Chr.11: 69005488-69005488 on Build GRCh38
rs570453145 Chr.11: 69005353-69005353 on Build GRCh38
rs148799278 Chr.11: 69005507-69005507 on Build GRCh38
rs745589039 Chr.11: 69005493-69005493 on Build GRCh38
rs763158831 Chr.11: 69005344-69005344 on Build GRCh38
rs959875761 Chr.11: 69005470-69005470 on Build GRCh38
rs867677887 Chr.11: 69005304-69005304 on Build GRCh38
rs762158049 Chr.11: 69005358-69005358 on Build GRCh38
rs199736282 Chr.11: 69005489-69005489 on Build GRCh38
rs778254527 Chr.11: 69005308-69005308 on Build GRCh38
rs755548309 Chr.11: 69005467-69005467 on Build GRCh38
rs11819876 Chr.11: 69005293-69005293 on Build GRCh38
rs766882878 Chr.11: 69005293-69005294 on Build GRCh38
rs779223706 Chr.11: 69005478-69005478 on Build GRCh38

View Full Product Details