Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 46907771-46908010 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MCFD2
Entrez Gene ID
90411
Gene Name
multiple coagulation factor deficiency 2
Gene Aliases
F5F8D, F5F8D2, LMAN1IP, SDNSF
UniGene
Hs.662152
Gene Chromosome Location
Chr.2: 46901870-46941855 on Build GRCh38
Transcripts
NM_001171506.2 NM_001171507.2 NM_001171508.2 NM_001171509.2 NM_001171510.2 NM_001171511.2 NM_139279.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3426483 c.203C>T p.S68L Chr.2: 46907916-46907916 on Build GRCh38
6273542 c.238C>A p.H80N Chr.2: 46907881-46907881 on Build GRCh38
5772552 c.227A>T p.Y76F Chr.2: 46907892-46907892 on Build GRCh38
1021212 c.204G>A p.S68S Chr.2: 46907915-46907915 on Build GRCh38
3839602 c.219G>A p.Q73Q Chr.2: 46907900-46907900 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763394078 Chr.2: 46907881-46907881 on Build GRCh38
rs777741288 Chr.2: 46907932-46907932 on Build GRCh38
rs754442571 Chr.2: 46907916-46907916 on Build GRCh38
rs747979718 Chr.2: 46907794-46907794 on Build GRCh38
rs759679044 Chr.2: 46907832-46907832 on Build GRCh38
rs764322613 Chr.2: 46907882-46907882 on Build GRCh38
rs387906287 Chr.2: 46907809-46907809 on Build GRCh38
rs746007921 Chr.2: 46907948-46907948 on Build GRCh38
rs771003004 Chr.2: 46907938-46907938 on Build GRCh38
rs1006795586 Chr.2: 46907800-46907800 on Build GRCh38
rs770123754 Chr.2: 46907835-46907835 on Build GRCh38
rs748439070 Chr.2: 46907808-46907809 on Build GRCh38
rs200415122 Chr.2: 46907925-46907925 on Build GRCh38
rs767997604 Chr.2: 46907905-46907905 on Build GRCh38
rs147861994 Chr.2: 46907891-46907891 on Build GRCh38
rs79211648 Chr.2: 46907820-46907820 on Build GRCh38
rs535874558 Chr.2: 46907914-46907914 on Build GRCh38
rs1017647378 Chr.2: 46907840-46907840 on Build GRCh38
rs78289603 Chr.2: 46907878-46907878 on Build GRCh38
rs994703366 Chr.2: 46907850-46907850 on Build GRCh38
rs774713036 Chr.2: 46907986-46907986 on Build GRCh38
rs759399774 Chr.2: 46907937-46907937 on Build GRCh38
rs751887064 Chr.2: 46907884-46907884 on Build GRCh38
rs746904307 Chr.2: 46907817-46907817 on Build GRCh38
rs752268860 Chr.2: 46907924-46907924 on Build GRCh38
rs543577609 Chr.2: 46907959-46907959 on Build GRCh38
rs137964402 Chr.2: 46907902-46907902 on Build GRCh38
rs192820723 Chr.2: 46907943-46907943 on Build GRCh38
rs777789932 Chr.2: 46907806-46907806 on Build GRCh38
rs775513362 Chr.2: 46907856-46907856 on Build GRCh38
rs914718297 Chr.2: 46907929-46907929 on Build GRCh38
rs144700781 Chr.2: 46907915-46907915 on Build GRCh38
rs966065013 Chr.2: 46907883-46907883 on Build GRCh38
rs763118443 Chr.2: 46907981-46907981 on Build GRCh38
rs773238774 Chr.2: 46907915-46907926 on Build GRCh38
rs768877565 Chr.2: 46907982-46907982 on Build GRCh38
rs770152708 Chr.2: 46907899-46907913 on Build GRCh38
rs771706958 Chr.2: 46907798-46907798 on Build GRCh38
rs767195582 Chr.2: 46907980-46907981 on Build GRCh38
rs138519672 Chr.2: 46907937-46907937 on Build GRCh38
rs558502042 Chr.2: 46907960-46907960 on Build GRCh38
rs886056118 Chr.2: 46907883-46907883 on Build GRCh38
rs775825745 Chr.2: 46907963-46907963 on Build GRCh38
rs778684574 Chr.2: 46907921-46907921 on Build GRCh38
rs776901670 Chr.2: 46907831-46907831 on Build GRCh38
rs146380249 Chr.2: 46907969-46907969 on Build GRCh38

View Full Product Details