Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 23529359-23529606 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EARS2
Entrez Gene ID
124454
Gene Name
glutamyl-tRNA synthetase 2, mitochondrial
Gene Aliases
COXPD12, MSE1, gluRS
UniGene
Hs.620541
Gene Chromosome Location
Chr.16: 23522013-23557375 on Build GRCh38
Transcripts
NM_001083614.1 NM_001308211.1 NR_003501.1 XM_011545738.1 XM_011545739.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3420864 c.1279C>T p.R427C Chr.16: 23529575-23529575 on Build GRCh38
5509516 c.1336G>A p.A446T Chr.16: 23529518-23529518 on Build GRCh38
6144087 c.1286C>T p.A429V Chr.16: 23529568-23529568 on Build GRCh38
1376796 c.1270C>A p.L424M Chr.16: 23529584-23529584 on Build GRCh38
3988348 c.1347G>A p.V449V Chr.16: 23529507-23529507 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs931216888 Chr.16: 23529438-23529438 on Build GRCh38
rs977767579 Chr.16: 23529575-23529575 on Build GRCh38
rs374785094 Chr.16: 23529567-23529567 on Build GRCh38
rs759251896 Chr.16: 23529580-23529580 on Build GRCh38
rs936524410 Chr.16: 23529409-23529409 on Build GRCh38
rs147414358 Chr.16: 23529487-23529487 on Build GRCh38
rs201181229 Chr.16: 23529502-23529502 on Build GRCh38
rs763412528 Chr.16: 23529495-23529495 on Build GRCh38
rs753414156 Chr.16: 23529574-23529575 on Build GRCh38
rs766085634 Chr.16: 23529497-23529497 on Build GRCh38
rs774926733 Chr.16: 23529577-23529577 on Build GRCh38
rs1011822758 Chr.16: 23529546-23529546 on Build GRCh38
rs74014932 Chr.16: 23529537-23529537 on Build GRCh38
rs781221918 Chr.16: 23529460-23529460 on Build GRCh38
rs773797383 Chr.16: 23529572-23529572 on Build GRCh38
rs752312159 Chr.16: 23529511-23529511 on Build GRCh38
rs764133947 Chr.16: 23529514-23529514 on Build GRCh38
rs756146515 Chr.16: 23529466-23529466 on Build GRCh38
rs747041821 Chr.16: 23529485-23529485 on Build GRCh38
rs777418791 Chr.16: 23529472-23529472 on Build GRCh38
rs959119286 Chr.16: 23529518-23529518 on Build GRCh38
rs777055582 Chr.16: 23529491-23529491 on Build GRCh38
rs200175968 Chr.16: 23529463-23529463 on Build GRCh38
rs770389020 Chr.16: 23529492-23529492 on Build GRCh38
rs757179196 Chr.16: 23529520-23529524 on Build GRCh38
rs748104284 Chr.16: 23529388-23529388 on Build GRCh38
rs755258609 Chr.16: 23529544-23529544 on Build GRCh38
rs1000267066 Chr.16: 23529434-23529434 on Build GRCh38
rs767133394 Chr.16: 23529503-23529503 on Build GRCh38
rs749562081 Chr.16: 23529574-23529574 on Build GRCh38
rs199853353 Chr.16: 23529512-23529512 on Build GRCh38
rs756327650 Chr.16: 23529428-23529428 on Build GRCh38
rs749158260 Chr.16: 23529474-23529474 on Build GRCh38
rs781117573 Chr.16: 23529545-23529545 on Build GRCh38
rs1013779280 Chr.16: 23529468-23529468 on Build GRCh38
rs766180648 Chr.16: 23529451-23529451 on Build GRCh38
rs201147154 Chr.16: 23529458-23529458 on Build GRCh38
rs375342248 Chr.16: 23529440-23529440 on Build GRCh38
rs531652112 Chr.16: 23529457-23529457 on Build GRCh38
rs1028937959 Chr.16: 23529559-23529559 on Build GRCh38
rs753537887 Chr.16: 23529516-23529516 on Build GRCh38
rs747235837 Chr.16: 23529538-23529538 on Build GRCh38
rs757066914 Chr.16: 23529526-23529526 on Build GRCh38
rs112604556 Chr.16: 23529562-23529562 on Build GRCh38
rs772276606 Chr.16: 23529420-23529420 on Build GRCh38
rs2072062 Chr.16: 23529477-23529477 on Build GRCh38
rs774164131 Chr.16: 23529500-23529500 on Build GRCh38

View Full Product Details