Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 44260748-44260997 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNMT3L
Entrez Gene ID
29947
Gene Name
DNA methyltransferase 3 like
Gene Aliases
-
UniGene
Hs.592165
Gene Chromosome Location
Chr.21: 44246339-44262216 on Build GRCh38
Transcripts
NM_013369.3 NM_175867.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4101919 c.139C>T p.R47* Chr.21: 44260807-44260807 on Build GRCh38
5942906 c.140G>A p.R47Q Chr.21: 44260806-44260806 on Build GRCh38
1031237 c.142A>G p.N48D Chr.21: 44260804-44260804 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs534646099 Chr.21: 44260837-44260837 on Build GRCh38
rs771333544 Chr.21: 44260823-44260823 on Build GRCh38
rs753815562 Chr.21: 44260790-44260790 on Build GRCh38
rs754950134 Chr.21: 44260873-44260873 on Build GRCh38
rs772926515 Chr.21: 44260853-44260853 on Build GRCh38
rs776822712 Chr.21: 44260833-44260833 on Build GRCh38
rs767496460 Chr.21: 44260832-44260833 on Build GRCh38
rs570968707 Chr.21: 44260966-44260966 on Build GRCh38
rs992205794 Chr.21: 44260962-44260962 on Build GRCh38
rs761710652 Chr.21: 44260805-44260806 on Build GRCh38
rs537252424 Chr.21: 44260768-44260768 on Build GRCh38
rs933908454 Chr.21: 44260769-44260769 on Build GRCh38
rs748199304 Chr.21: 44260799-44260799 on Build GRCh38
rs758691224 Chr.21: 44260801-44260801 on Build GRCh38
rs114308751 Chr.21: 44260925-44260925 on Build GRCh38
rs570957584 Chr.21: 44260863-44260863 on Build GRCh38
rs755921084 Chr.21: 44260771-44260771 on Build GRCh38
rs370648505 Chr.21: 44260782-44260782 on Build GRCh38
rs374776676 Chr.21: 44260797-44260797 on Build GRCh38
rs755119167 Chr.21: 44260965-44260965 on Build GRCh38
rs373680073 Chr.21: 44260849-44260849 on Build GRCh38
rs763913786 Chr.21: 44260800-44260800 on Build GRCh38
rs1033844785 Chr.21: 44260909-44260909 on Build GRCh38
rs951020003 Chr.21: 44260944-44260944 on Build GRCh38
rs956995780 Chr.21: 44260930-44260930 on Build GRCh38
rs750174301 Chr.21: 44260841-44260841 on Build GRCh38
rs761403685 Chr.21: 44260848-44260848 on Build GRCh38
rs72613643 Chr.21: 44260899-44260899 on Build GRCh38
rs765230620 Chr.21: 44260924-44260924 on Build GRCh38
rs755186540 Chr.21: 44260950-44260950 on Build GRCh38
rs201320726 Chr.21: 44260806-44260806 on Build GRCh38
rs775503239 Chr.21: 44260788-44260791 on Build GRCh38
rs368058982 Chr.21: 44260807-44260807 on Build GRCh38
rs368717133 Chr.21: 44260864-44260864 on Build GRCh38
rs765158396 Chr.21: 44260888-44260888 on Build GRCh38
rs370267082 Chr.21: 44260845-44260845 on Build GRCh38
rs753555640 Chr.21: 44260871-44260871 on Build GRCh38
rs553210775 Chr.21: 44260841-44260841 on Build GRCh38
rs754864909 Chr.21: 44260793-44260793 on Build GRCh38

View Full Product Details