Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 27890203-27890476 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIST1H3J
Entrez Gene ID
8356
Gene Name
histone cluster 1, H3j
Gene Aliases
H3/j, H3FJ
UniGene
Hs.484990
Gene Chromosome Location
Chr.6: 27890315-27890792 on Build GRCh38
Transcripts
NM_003535.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1076986 c.386G>A p.R129H Chr.6: 27890407-27890407 on Build GRCh38
6987274 c.378G>C p.Q126H Chr.6: 27890415-27890415 on Build GRCh38
3249572 c.349C>T p.R117C Chr.6: 27890444-27890444 on Build GRCh38
5948854 c.403C>G p.R135G Chr.6: 27890390-27890390 on Build GRCh38
6939856 c.385C>T p.R129C Chr.6: 27890408-27890408 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs916613042 Chr.6: 27890421-27890421 on Build GRCh38
rs750516025 Chr.6: 27890351-27890351 on Build GRCh38
rs758449003 Chr.6: 27890372-27890372 on Build GRCh38
rs961945297 Chr.6: 27890343-27890343 on Build GRCh38
rs766187270 Chr.6: 27890337-27890337 on Build GRCh38
rs775659971 Chr.6: 27890451-27890451 on Build GRCh38
rs759804139 Chr.6: 27890356-27890356 on Build GRCh38
rs759280383 Chr.6: 27890443-27890443 on Build GRCh38
rs953144491 Chr.6: 27890274-27890274 on Build GRCh38
rs772995711 Chr.6: 27890336-27890336 on Build GRCh38
rs773173554 Chr.6: 27890331-27890331 on Build GRCh38
rs757621757 Chr.6: 27890378-27890378 on Build GRCh38
rs986311776 Chr.6: 27890288-27890289 on Build GRCh38
rs747698805 Chr.6: 27890409-27890409 on Build GRCh38
rs61734000 Chr.6: 27890414-27890414 on Build GRCh38
rs746976536 Chr.6: 27890427-27890427 on Build GRCh38
rs746178535 Chr.6: 27890381-27890381 on Build GRCh38
rs758616223 Chr.6: 27890369-27890369 on Build GRCh38
rs781623203 Chr.6: 27890379-27890379 on Build GRCh38
rs765494141 Chr.6: 27890446-27890446 on Build GRCh38
rs765401144 Chr.6: 27890357-27890357 on Build GRCh38
rs202049846 Chr.6: 27890368-27890368 on Build GRCh38
rs373809153 Chr.6: 27890345-27890352 on Build GRCh38
rs997740664 Chr.6: 27890256-27890256 on Build GRCh38
rs768443928 Chr.6: 27890385-27890385 on Build GRCh38
rs184135815 Chr.6: 27890268-27890268 on Build GRCh38
rs770711648 Chr.6: 27890431-27890431 on Build GRCh38
rs531357806 Chr.6: 27890302-27890302 on Build GRCh38
rs533352985 Chr.6: 27890257-27890257 on Build GRCh38
rs771701828 Chr.6: 27890411-27890411 on Build GRCh38
rs778684793 Chr.6: 27890394-27890394 on Build GRCh38
rs775919880 Chr.6: 27890337-27890344 on Build GRCh38
rs776524828 Chr.6: 27890432-27890432 on Build GRCh38
rs1005998933 Chr.6: 27890326-27890326 on Build GRCh38
rs922296721 Chr.6: 27890418-27890418 on Build GRCh38
rs948039279 Chr.6: 27890426-27890426 on Build GRCh38
rs73386651 Chr.6: 27890262-27890262 on Build GRCh38
rs752103654 Chr.6: 27890376-27890376 on Build GRCh38
rs553160475 Chr.6: 27890314-27890315 on Build GRCh38
rs373448729 Chr.6: 27890307-27890308 on Build GRCh38
rs757487872 Chr.6: 27890289-27890289 on Build GRCh38
rs201216167 Chr.6: 27890373-27890373 on Build GRCh38
rs968503610 Chr.6: 27890276-27890276 on Build GRCh38
rs928439781 Chr.6: 27890364-27890364 on Build GRCh38

View Full Product Details