Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 51345162-51345419 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ETFB
Entrez Gene ID
2109
Gene Name
electron transfer flavoprotein beta subunit
Gene Aliases
FP585, MADD
UniGene
Hs.348531
Gene Chromosome Location
Chr.19: 51345155-51366418 on Build GRCh38
Transcripts
NM_001014763.1 NM_001985.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2755991 c.649G>T p.D217Y Chr.19: 51345330-51345330 on Build GRCh38
6857918 c.891C>G p.I297M Chr.19: 51345361-51345361 on Build GRCh38
5056876 c.894G>T p.E298D Chr.19: 51345358-51345358 on Build GRCh38
6897655 c.924C>T p.D308D Chr.19: 51345328-51345328 on Build GRCh38
1751100 c.996C>T p.T332T Chr.19: 51345256-51345256 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771560927 Chr.19: 51345327-51345327 on Build GRCh38
rs11559094 Chr.19: 51345357-51345357 on Build GRCh38
rs375326450 Chr.19: 51345273-51345273 on Build GRCh38
rs767519060 Chr.19: 51345363-51345365 on Build GRCh38
rs879899472 Chr.19: 51345343-51345343 on Build GRCh38
rs763960961 Chr.19: 51345206-51345206 on Build GRCh38
rs779532662 Chr.19: 51345330-51345330 on Build GRCh38
rs548267062 Chr.19: 51345297-51345297 on Build GRCh38
rs983860068 Chr.19: 51345389-51345389 on Build GRCh38
rs749837573 Chr.19: 51345271-51345271 on Build GRCh38
rs746450125 Chr.19: 51345232-51345232 on Build GRCh38
rs769668781 Chr.19: 51345277-51345277 on Build GRCh38
rs746160252 Chr.19: 51345333-51345333 on Build GRCh38
rs775793200 Chr.19: 51345276-51345276 on Build GRCh38
rs149129214 Chr.19: 51345270-51345270 on Build GRCh38
rs761714465 Chr.19: 51345282-51345282 on Build GRCh38
rs780828891 Chr.19: 51345316-51345316 on Build GRCh38
rs765255418 Chr.19: 51345289-51345289 on Build GRCh38
rs757003787 Chr.19: 51345209-51345209 on Build GRCh38
rs200877363 Chr.19: 51345292-51345292 on Build GRCh38
rs145010124 Chr.19: 51345290-51345290 on Build GRCh38
rs775532173 Chr.19: 51345344-51345344 on Build GRCh38
rs145173884 Chr.19: 51345218-51345218 on Build GRCh38
rs768290418 Chr.19: 51345265-51345265 on Build GRCh38
rs370522844 Chr.19: 51345219-51345219 on Build GRCh38
rs768912021 Chr.19: 51345353-51345353 on Build GRCh38
rs756243839 Chr.19: 51345195-51345195 on Build GRCh38
rs747702919 Chr.19: 51345317-51345317 on Build GRCh38
rs139315136 Chr.19: 51345313-51345313 on Build GRCh38
rs779819237 Chr.19: 51345229-51345229 on Build GRCh38
rs760012472 Chr.19: 51345274-51345274 on Build GRCh38
rs776658637 Chr.19: 51345360-51345360 on Build GRCh38
rs398123677 Chr.19: 51345251-51345251 on Build GRCh38
rs533794107 Chr.19: 51345361-51345361 on Build GRCh38
rs772489973 Chr.19: 51345338-51345338 on Build GRCh38
rs750961823 Chr.19: 51345382-51345382 on Build GRCh38
rs765068707 Chr.19: 51345376-51345376 on Build GRCh38
rs753778245 Chr.19: 51345207-51345207 on Build GRCh38
rs760907327 Chr.19: 51345347-51345347 on Build GRCh38
rs952095878 Chr.19: 51345382-51345382 on Build GRCh38
rs920672041 Chr.19: 51345381-51345381 on Build GRCh38
rs764188419 Chr.19: 51345278-51345278 on Build GRCh38
rs750381743 Chr.19: 51345395-51345395 on Build GRCh38
rs761894535 Chr.19: 51345264-51345264 on Build GRCh38
rs529795334 Chr.19: 51345281-51345281 on Build GRCh38

View Full Product Details