Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 73232243-73232502 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf80
Entrez Gene ID
55028
Gene Name
chromosome 17 open reading frame 80
Gene Aliases
HLC-8, MIG3, SPEP1
UniGene
Hs.12929
Gene Chromosome Location
Chr.17: 73232233-73248959 on Build GRCh38
Transcripts
NM_001288770.1 NM_001288771.1 XM_005257487.3 NM_001098832.1 NM_001289410.1 NM_001289411.1 NM_032837.2

Gene Symbol
FAM104A
Entrez Gene ID
84923
Gene Name
family with sequence similarity 104 member A
Gene Aliases
-
UniGene
Hs.741276
Gene Chromosome Location
Chr.17: 73207353-73232394 on Build GRCh38
Transcripts
NM_001288770.1 NM_001288771.1 XM_005257487.3 NM_001098832.1 NM_001289410.1 NM_001289411.1 NM_032837.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3521590 c.58T>A p.Y20N Chr.17: 73232249-73232249 on Build GRCh38
6556628 c.2T>A p.M1K Chr.17: 73232305-73232305 on Build GRCh38
7109473 c.15_25del11 p.A6fs*2 Chr.17: 73232282-73232292 on Build GRCh38
4614394 c.9delG p.R4fs*64 Chr.17: 73232298-73232298 on Build GRCh38
359133 c.52G>A p.E18K Chr.17: 73232255-73232255 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs959697233 Chr.17: 73232423-73232423 on Build GRCh38
rs773493866 Chr.17: 73232298-73232298 on Build GRCh38
rs375693359 Chr.17: 73232337-73232337 on Build GRCh38
rs544913626 Chr.17: 73232359-73232359 on Build GRCh38
rs781642632 Chr.17: 73232263-73232263 on Build GRCh38
rs770953647 Chr.17: 73232332-73232333 on Build GRCh38
rs761753641 Chr.17: 73232329-73232329 on Build GRCh38
rs770000277 Chr.17: 73232297-73232298 on Build GRCh38
rs755310458 Chr.17: 73232260-73232260 on Build GRCh38
rs941287240 Chr.17: 73232291-73232291 on Build GRCh38
rs988568802 Chr.17: 73232306-73232306 on Build GRCh38
rs775222018 Chr.17: 73232284-73232284 on Build GRCh38
rs770422960 Chr.17: 73232274-73232274 on Build GRCh38
rs1035237612 Chr.17: 73232419-73232419 on Build GRCh38
rs745410064 Chr.17: 73232312-73232312 on Build GRCh38
rs377066963 Chr.17: 73232363-73232363 on Build GRCh38
rs1003811928 Chr.17: 73232407-73232407 on Build GRCh38
rs377592119 Chr.17: 73232304-73232304 on Build GRCh38
rs755185778 Chr.17: 73232302-73232302 on Build GRCh38
rs76917873 Chr.17: 73232479-73232479 on Build GRCh38
rs949929969 Chr.17: 73232447-73232447 on Build GRCh38
rs748324805 Chr.17: 73232269-73232269 on Build GRCh38
rs773744672 Chr.17: 73232265-73232265 on Build GRCh38
rs751651372 Chr.17: 73232336-73232336 on Build GRCh38
rs775189353 Chr.17: 73232319-73232319 on Build GRCh38
rs548215808 Chr.17: 73232300-73232300 on Build GRCh38
rs560347523 Chr.17: 73232361-73232361 on Build GRCh38
rs899242254 Chr.17: 73232366-73232366 on Build GRCh38
rs751702545 Chr.17: 73232299-73232299 on Build GRCh38
rs370554488 Chr.17: 73232309-73232309 on Build GRCh38
rs764479072 Chr.17: 73232464-73232464 on Build GRCh38
rs969689298 Chr.17: 73232404-73232404 on Build GRCh38
rs373736498 Chr.17: 73232313-73232313 on Build GRCh38
rs995050294 Chr.17: 73232369-73232369 on Build GRCh38
rs909878911 Chr.17: 73232416-73232416 on Build GRCh38
rs367575990 Chr.17: 73232327-73232327 on Build GRCh38
rs761727566 Chr.17: 73232296-73232296 on Build GRCh38
rs941411988 Chr.17: 73232430-73232430 on Build GRCh38
rs200728518 Chr.17: 73232322-73232322 on Build GRCh38
rs544578624 Chr.17: 73232303-73232303 on Build GRCh38
rs760809940 Chr.17: 73232444-73232445 on Build GRCh38
rs753786628 Chr.17: 73232293-73232293 on Build GRCh38
rs970035327 Chr.17: 73232294-73232294 on Build GRCh38
rs760537927 Chr.17: 73232286-73232286 on Build GRCh38
rs549066550 Chr.17: 73232396-73232396 on Build GRCh38
rs753994928 Chr.17: 73232480-73232480 on Build GRCh38
rs767157414 Chr.17: 73232282-73232282 on Build GRCh38
rs201327693 Chr.17: 73232298-73232298 on Build GRCh38
rs199818438 Chr.17: 73232261-73232261 on Build GRCh38
rs749798858 Chr.17: 73232266-73232266 on Build GRCh38
rs781506110 Chr.17: 73232262-73232270 on Build GRCh38
rs763111693 Chr.17: 73232322-73232323 on Build GRCh38
rs1051606230 Chr.17: 73232355-73232355 on Build GRCh38
rs527407008 Chr.17: 73232368-73232368 on Build GRCh38
rs988474005 Chr.17: 73232445-73232445 on Build GRCh38
rs201703597 Chr.17: 73232331-73232331 on Build GRCh38
rs759126875 Chr.17: 73232279-73232279 on Build GRCh38

View Full Product Details