Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 139618774-139619023 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNK9
Entrez Gene ID
51305
Gene Name
potassium two pore domain channel subfamily K member 9
Gene Aliases
K2p9.1, KT3.2, TASK-3, TASK3
UniGene
Hs.493037
Gene Chromosome Location
Chr.8: 139600838-139703135 on Build GRCh38
Transcripts
NM_001282534.1 NR_104210.1 XM_011517101.2 XM_011517102.2 XM_011517103.2 XM_017013530.1 XM_017013531.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1489047 c.505G>A p.G169R Chr.8: 139618878-139618878 on Build GRCh38
5727718 c.432G>A p.K144K Chr.8: 139618951-139618951 on Build GRCh38
749034 c.500G>C p.C167S Chr.8: 139618883-139618883 on Build GRCh38
3645703 c.519C>T p.I173I Chr.8: 139618864-139618864 on Build GRCh38
4006856 c.401C>A p.T134N Chr.8: 139618982-139618982 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145617381 Chr.8: 139618867-139618867 on Build GRCh38
rs558869834 Chr.8: 139618978-139618978 on Build GRCh38
rs763947917 Chr.8: 139618974-139618974 on Build GRCh38
rs752611105 Chr.8: 139618796-139618796 on Build GRCh38
rs890247654 Chr.8: 139618936-139618936 on Build GRCh38
rs149575059 Chr.8: 139618951-139618951 on Build GRCh38
rs868757268 Chr.8: 139618846-139618846 on Build GRCh38
rs578106232 Chr.8: 139618996-139618996 on Build GRCh38
rs776045672 Chr.8: 139618924-139618924 on Build GRCh38
rs753123156 Chr.8: 139618959-139618959 on Build GRCh38
rs774431836 Chr.8: 139618935-139618935 on Build GRCh38
rs780709164 Chr.8: 139618873-139618873 on Build GRCh38
rs749780302 Chr.8: 139618925-139618925 on Build GRCh38
rs866795921 Chr.8: 139618856-139618856 on Build GRCh38
rs192943243 Chr.8: 139618852-139618852 on Build GRCh38
rs758314044 Chr.8: 139618973-139618973 on Build GRCh38
rs866426094 Chr.8: 139618849-139618849 on Build GRCh38
rs751012938 Chr.8: 139618977-139618977 on Build GRCh38
rs770303698 Chr.8: 139618885-139618885 on Build GRCh38
rs780846538 Chr.8: 139618993-139618993 on Build GRCh38
rs777257931 Chr.8: 139618855-139618855 on Build GRCh38
rs769346588 Chr.8: 139618934-139618934 on Build GRCh38
rs1008023884 Chr.8: 139619002-139619002 on Build GRCh38
rs373908677 Chr.8: 139618872-139618872 on Build GRCh38
rs868526327 Chr.8: 139618995-139618995 on Build GRCh38
rs900238044 Chr.8: 139618870-139618870 on Build GRCh38
rs767740721 Chr.8: 139618939-139618939 on Build GRCh38
rs565355081 Chr.8: 139618971-139618971 on Build GRCh38
rs746554309 Chr.8: 139618858-139618858 on Build GRCh38
rs867543866 Chr.8: 139618991-139618991 on Build GRCh38
rs758375356 Chr.8: 139618801-139618801 on Build GRCh38
rs898438493 Chr.8: 139618877-139618877 on Build GRCh38
rs764990488 Chr.8: 139618961-139618961 on Build GRCh38
rs768019389 Chr.8: 139618958-139618958 on Build GRCh38
rs995818177 Chr.8: 139618900-139618900 on Build GRCh38

View Full Product Details