Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 11765596-11765835 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MSL3
Entrez Gene ID
10943
Gene Name
male-specific lethal 3 homolog (Drosophila)
Gene Aliases
MSL3L1
UniGene
Hs.655288
Gene Chromosome Location
Chr.X: 11758159-11775753 on Build GRCh38
Transcripts
NM_001193270.2 NM_001282174.1 NM_006800.3 NM_078628.1 NM_078629.3 XM_005274440.2 XM_011545442.2

SNP Detail

SNP ID Chromosome Location
rs200140696 Chr.X: 11765638-11765638 on Build GRCh38
rs147621824 Chr.X: 11765758-11765758 on Build GRCh38
rs749880396 Chr.X: 11765793-11765793 on Build GRCh38
rs770246787 Chr.X: 11765649-11765649 on Build GRCh38
rs751079550 Chr.X: 11765654-11765654 on Build GRCh38
rs375933028 Chr.X: 11765694-11765694 on Build GRCh38
rs772470103 Chr.X: 11765743-11765743 on Build GRCh38
rs748332877 Chr.X: 11765768-11765768 on Build GRCh38
rs371263192 Chr.X: 11765685-11765685 on Build GRCh38
rs147804390 Chr.X: 11765652-11765652 on Build GRCh38
rs762239535 Chr.X: 11765642-11765642 on Build GRCh38
rs375478362 Chr.X: 11765802-11765802 on Build GRCh38
rs747197871 Chr.X: 11765617-11765617 on Build GRCh38
rs778218411 Chr.X: 11765753-11765753 on Build GRCh38
rs780120545 Chr.X: 11765615-11765615 on Build GRCh38
rs749183286 Chr.X: 11765613-11765613 on Build GRCh38
rs368501339 Chr.X: 11765757-11765757 on Build GRCh38
rs940265368 Chr.X: 11765689-11765689 on Build GRCh38
rs201837787 Chr.X: 11765800-11765800 on Build GRCh38
rs367610679 Chr.X: 11765683-11765683 on Build GRCh38
rs1002328169 Chr.X: 11765696-11765696 on Build GRCh38
rs372545766 Chr.X: 11765775-11765775 on Build GRCh38
rs1047921779 Chr.X: 11765810-11765810 on Build GRCh38
rs142090839 Chr.X: 11765614-11765614 on Build GRCh38
rs770295933 Chr.X: 11765771-11765771 on Build GRCh38
rs141301691 Chr.X: 11765653-11765653 on Build GRCh38
rs753786365 Chr.X: 11765706-11765706 on Build GRCh38
rs975529560 Chr.X: 11765644-11765644 on Build GRCh38
rs776536449 Chr.X: 11765667-11765667 on Build GRCh38
rs145217094 Chr.X: 11765692-11765692 on Build GRCh38
rs780063099 Chr.X: 11765750-11765750 on Build GRCh38
rs1011300169 Chr.X: 11765754-11765754 on Build GRCh38
rs768916433 Chr.X: 11765631-11765631 on Build GRCh38
rs773784866 Chr.X: 11765806-11765806 on Build GRCh38

View Full Product Details