Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 69419340-69419595 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MARVELD2
Entrez Gene ID
153562
Gene Name
MARVEL domain containing 2
Gene Aliases
DFNB49, MARVD2, MRVLDC2, Tric
UniGene
Hs.657687
Gene Chromosome Location
Chr.5: 69415112-69444022 on Build GRCh38
Transcripts
NM_001038603.2 NM_001244734.1 XM_005248445.4 XM_005248446.4 XM_005248447.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5868758 c.77C>T p.T26I Chr.5: 69419462-69419462 on Build GRCh38
7163407 c.87A>G p.R29R Chr.5: 69419472-69419472 on Build GRCh38
3856116 c.28C>T p.R10W Chr.5: 69419413-69419413 on Build GRCh38
3728584 c.78C>A p.T26T Chr.5: 69419463-69419463 on Build GRCh38
5975661 c.146C>T p.P49L Chr.5: 69419531-69419531 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138816138 Chr.5: 69419541-69419541 on Build GRCh38
rs376484431 Chr.5: 69419402-69419402 on Build GRCh38
rs190826169 Chr.5: 69419515-69419515 on Build GRCh38
rs888249127 Chr.5: 69419453-69419453 on Build GRCh38
rs749198352 Chr.5: 69419516-69419516 on Build GRCh38
rs776724674 Chr.5: 69419568-69419568 on Build GRCh38
rs150434290 Chr.5: 69419561-69419561 on Build GRCh38
rs780599259 Chr.5: 69419512-69419512 on Build GRCh38
rs142002229 Chr.5: 69419556-69419556 on Build GRCh38
rs751155210 Chr.5: 69419412-69419412 on Build GRCh38
rs367912254 Chr.5: 69419539-69419539 on Build GRCh38
rs745712719 Chr.5: 69419443-69419443 on Build GRCh38
rs762606820 Chr.5: 69419406-69419406 on Build GRCh38
rs778447160 Chr.5: 69419569-69419578 on Build GRCh38
rs1185246 Chr.5: 69419483-69419483 on Build GRCh38
rs768778743 Chr.5: 69419467-69419467 on Build GRCh38
rs200008142 Chr.5: 69419414-69419414 on Build GRCh38
rs763810648 Chr.5: 69419524-69419553 on Build GRCh38
rs748212061 Chr.5: 69419503-69419503 on Build GRCh38
rs531073647 Chr.5: 69419573-69419573 on Build GRCh38
rs755442661 Chr.5: 69419550-69419550 on Build GRCh38
rs199777123 Chr.5: 69419422-69419422 on Build GRCh38
rs765300957 Chr.5: 69419392-69419392 on Build GRCh38
rs555221544 Chr.5: 69419449-69419449 on Build GRCh38
rs766746155 Chr.5: 69419492-69419492 on Build GRCh38
rs978969615 Chr.5: 69419390-69419390 on Build GRCh38
rs111458976 Chr.5: 69419571-69419571 on Build GRCh38
rs752493567 Chr.5: 69419502-69419502 on Build GRCh38
rs761504482 Chr.5: 69419479-69419479 on Build GRCh38
rs374313745 Chr.5: 69419423-69419423 on Build GRCh38
rs759473286 Chr.5: 69419373-69419373 on Build GRCh38
rs763656680 Chr.5: 69419470-69419470 on Build GRCh38
rs540714945 Chr.5: 69419491-69419491 on Build GRCh38
rs748364395 Chr.5: 69419428-69419428 on Build GRCh38
rs373371842 Chr.5: 69419371-69419371 on Build GRCh38
rs777924191 Chr.5: 69419563-69419563 on Build GRCh38
rs772495236 Chr.5: 69419375-69419375 on Build GRCh38
rs145027254 Chr.5: 69419500-69419500 on Build GRCh38
rs773755999 Chr.5: 69419475-69419475 on Build GRCh38
rs768754255 Chr.5: 69419518-69419518 on Build GRCh38
rs761286508 Chr.5: 69419521-69419521 on Build GRCh38
rs544651660 Chr.5: 69419532-69419532 on Build GRCh38
rs752821193 Chr.5: 69419557-69419557 on Build GRCh38
rs777927237 Chr.5: 69419511-69419511 on Build GRCh38
rs760474616 Chr.5: 69419519-69419520 on Build GRCh38
rs112747252 Chr.5: 69419416-69419416 on Build GRCh38
rs762986895 Chr.5: 69419469-69419469 on Build GRCh38
rs756888886 Chr.5: 69419413-69419413 on Build GRCh38
rs754228965 Chr.5: 69419545-69419545 on Build GRCh38
rs750796880 Chr.5: 69419485-69419485 on Build GRCh38
rs778971496 Chr.5: 69419427-69419427 on Build GRCh38
rs756855195 Chr.5: 69419565-69419565 on Build GRCh38
rs769838733 Chr.5: 69419446-69419446 on Build GRCh38
rs755495483 Chr.5: 69419501-69419501 on Build GRCh38
rs753451291 Chr.5: 69419559-69419561 on Build GRCh38
rs772209957 Chr.5: 69419442-69419442 on Build GRCh38
rs143592561 Chr.5: 69419415-69419415 on Build GRCh38
rs752477482 Chr.5: 69419511-69419513 on Build GRCh38

View Full Product Details