Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 36479456-36479729 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRIM1
Entrez Gene ID
51232
Gene Name
cysteine rich transmembrane BMP regulator 1
Gene Aliases
CRIM-1, S52
UniGene
Hs.699247
Gene Chromosome Location
Chr.2: 36356088-36551135 on Build GRCh38
Transcripts
NM_016441.2 XM_005264357.4 XM_011532898.2 XM_011532899.2 XM_011532901.2 XM_017004258.1 XM_017004259.1 XM_017004260.1 XM_017004261.1 XM_017004262.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
265688 c.1250G>A p.R417Q Chr.2: 36479572-36479572 on Build GRCh38
327403 c.1248G>A p.W416* Chr.2: 36479570-36479570 on Build GRCh38
367239 c.1284C>T p.N428N Chr.2: 36479606-36479606 on Build GRCh38
6542520 c.1340A>T p.K447I Chr.2: 36479662-36479662 on Build GRCh38
107114 c.1187C>T p.P396L Chr.2: 36479509-36479509 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751158790 Chr.2: 36479683-36479683 on Build GRCh38
rs371942777 Chr.2: 36479600-36479600 on Build GRCh38
rs202040233 Chr.2: 36479690-36479690 on Build GRCh38
rs750151277 Chr.2: 36479671-36479671 on Build GRCh38
rs147700058 Chr.2: 36479627-36479627 on Build GRCh38
rs551904478 Chr.2: 36479483-36479483 on Build GRCh38
rs779719906 Chr.2: 36479626-36479626 on Build GRCh38
rs746293769 Chr.2: 36479637-36479637 on Build GRCh38
rs138940127 Chr.2: 36479645-36479645 on Build GRCh38
rs766800438 Chr.2: 36479505-36479505 on Build GRCh38
rs77413996 Chr.2: 36479687-36479687 on Build GRCh38
rs760795693 Chr.2: 36479580-36479580 on Build GRCh38
rs750287447 Chr.2: 36479615-36479615 on Build GRCh38
rs767565422 Chr.2: 36479572-36479572 on Build GRCh38
rs779414943 Chr.2: 36479545-36479545 on Build GRCh38
rs940916186 Chr.2: 36479516-36479516 on Build GRCh38
rs757223335 Chr.2: 36479607-36479607 on Build GRCh38
rs752849582 Chr.2: 36479606-36479606 on Build GRCh38
rs920875854 Chr.2: 36479666-36479666 on Build GRCh38
rs7574705 Chr.2: 36479522-36479522 on Build GRCh38
rs771535773 Chr.2: 36479631-36479631 on Build GRCh38
rs774391886 Chr.2: 36479506-36479506 on Build GRCh38
rs531660918 Chr.2: 36479501-36479501 on Build GRCh38
rs140732031 Chr.2: 36479622-36479622 on Build GRCh38
rs139895660 Chr.2: 36479539-36479539 on Build GRCh38
rs768788721 Chr.2: 36479655-36479655 on Build GRCh38
rs899467815 Chr.2: 36479614-36479614 on Build GRCh38
rs998744411 Chr.2: 36479613-36479613 on Build GRCh38
rs760990343 Chr.2: 36479654-36479654 on Build GRCh38
rs375085387 Chr.2: 36479660-36479660 on Build GRCh38
rs369088958 Chr.2: 36479554-36479554 on Build GRCh38
rs572243696 Chr.2: 36479559-36479559 on Build GRCh38
rs763350869 Chr.2: 36479503-36479503 on Build GRCh38
rs570331773 Chr.2: 36479499-36479499 on Build GRCh38
rs764933162 Chr.2: 36479610-36479610 on Build GRCh38
rs142581295 Chr.2: 36479558-36479558 on Build GRCh38
rs141894588 Chr.2: 36479691-36479691 on Build GRCh38
rs766187610 Chr.2: 36479682-36479682 on Build GRCh38
rs188554284 Chr.2: 36479528-36479528 on Build GRCh38
rs202222795 Chr.2: 36479565-36479565 on Build GRCh38
rs759674160 Chr.2: 36479566-36479566 on Build GRCh38
rs146435632 Chr.2: 36479636-36479636 on Build GRCh38
rs775804277 Chr.2: 36479649-36479649 on Build GRCh38
rs764178371 Chr.2: 36479523-36479523 on Build GRCh38
rs541765928 Chr.2: 36479662-36479662 on Build GRCh38
rs143510576 Chr.2: 36479509-36479509 on Build GRCh38
rs576743714 Chr.2: 36479629-36479629 on Build GRCh38
rs77551206 Chr.2: 36479630-36479630 on Build GRCh38
rs113389944 Chr.2: 36479706-36479706 on Build GRCh38
rs941069110 Chr.2: 36479591-36479591 on Build GRCh38
rs576125560 Chr.2: 36479663-36479663 on Build GRCh38
rs141419153 Chr.2: 36479658-36479658 on Build GRCh38
rs146015641 Chr.2: 36479678-36479678 on Build GRCh38
rs113372122 Chr.2: 36479557-36479557 on Build GRCh38
rs550286125 Chr.2: 36479502-36479502 on Build GRCh38
rs1032947970 Chr.2: 36479616-36479616 on Build GRCh38
rs1045365045 Chr.2: 36479571-36479571 on Build GRCh38
rs775691814 Chr.2: 36479579-36479579 on Build GRCh38
rs778782436 Chr.2: 36479536-36479536 on Build GRCh38

View Full Product Details