Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 43675107-43675380 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MRPS18A
Entrez Gene ID
55168
Gene Name
mitochondrial ribosomal protein S18A
Gene Aliases
HumanS18b, MRP-S18-3, MRPS18-3, S18bmt
UniGene
Hs.520149
Gene Chromosome Location
Chr.6: 43671197-43687812 on Build GRCh38
Transcripts
NM_001193343.1 NM_018135.3 XM_006715134.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6380930 c.668G>C p.R223P Chr.6: 43675202-43675202 on Build GRCh38
1444771 c.422C>T p.P141L Chr.6: 43675226-43675226 on Build GRCh38
6587975 c.648G>C p.K216N Chr.6: 43675222-43675222 on Build GRCh38
4446578 c.535C>T p.P179S Chr.6: 43675335-43675335 on Build GRCh38
1257952 c.377-8C>T p.? Chr.6: 43675279-43675279 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs573042146 Chr.6: 43675290-43675290 on Build GRCh38
rs759452064 Chr.6: 43675167-43675167 on Build GRCh38
rs766307881 Chr.6: 43675292-43675292 on Build GRCh38
rs140706518 Chr.6: 43675210-43675210 on Build GRCh38
rs267601047 Chr.6: 43675284-43675284 on Build GRCh38
rs775568128 Chr.6: 43675326-43675326 on Build GRCh38
rs544543545 Chr.6: 43675350-43675350 on Build GRCh38
rs753835432 Chr.6: 43675165-43675165 on Build GRCh38
rs149334831 Chr.6: 43675247-43675247 on Build GRCh38
rs775235245 Chr.6: 43675232-43675232 on Build GRCh38
rs567137815 Chr.6: 43675299-43675299 on Build GRCh38
rs769201835 Chr.6: 43675152-43675152 on Build GRCh38
rs754376107 Chr.6: 43675202-43675202 on Build GRCh38
rs772265293 Chr.6: 43675213-43675213 on Build GRCh38
rs528290524 Chr.6: 43675203-43675203 on Build GRCh38
rs757560907 Chr.6: 43675298-43675299 on Build GRCh38
rs563108824 Chr.6: 43675143-43675143 on Build GRCh38
rs201797896 Chr.6: 43675276-43675276 on Build GRCh38
rs202096992 Chr.6: 43675267-43675267 on Build GRCh38
rs201676131 Chr.6: 43675163-43675163 on Build GRCh38
rs747195773 Chr.6: 43675257-43675275 on Build GRCh38
rs758864098 Chr.6: 43675283-43675283 on Build GRCh38
rs772368068 Chr.6: 43675157-43675157 on Build GRCh38
rs371004644 Chr.6: 43675219-43675219 on Build GRCh38
rs774836778 Chr.6: 43675153-43675153 on Build GRCh38
rs752504057 Chr.6: 43675171-43675171 on Build GRCh38
rs990545599 Chr.6: 43675144-43675144 on Build GRCh38
rs761983423 Chr.6: 43675154-43675154 on Build GRCh38
rs749986587 Chr.6: 43675271-43675271 on Build GRCh38
rs777497866 Chr.6: 43675175-43675175 on Build GRCh38
rs200075534 Chr.6: 43675252-43675252 on Build GRCh38
rs773486967 Chr.6: 43675214-43675214 on Build GRCh38
rs377621054 Chr.6: 43675192-43675192 on Build GRCh38
rs777982223 Chr.6: 43675285-43675285 on Build GRCh38
rs377074058 Chr.6: 43675162-43675162 on Build GRCh38
rs571547048 Chr.6: 43675225-43675225 on Build GRCh38
rs767302785 Chr.6: 43675351-43675351 on Build GRCh38
rs968291037 Chr.6: 43675188-43675188 on Build GRCh38
rs373418874 Chr.6: 43675170-43675170 on Build GRCh38
rs769801553 Chr.6: 43675318-43675318 on Build GRCh38
rs371619512 Chr.6: 43675282-43675282 on Build GRCh38
rs771738289 Chr.6: 43675352-43675352 on Build GRCh38
rs557240042 Chr.6: 43675248-43675248 on Build GRCh38
rs558754453 Chr.6: 43675308-43675308 on Build GRCh38
rs200771687 Chr.6: 43675195-43675195 on Build GRCh38
rs751110566 Chr.6: 43675187-43675187 on Build GRCh38
rs200253909 Chr.6: 43675342-43675342 on Build GRCh38
rs766379790 Chr.6: 43675164-43675164 on Build GRCh38
rs777667446 Chr.6: 43675166-43675166 on Build GRCh38
rs975727289 Chr.6: 43675293-43675293 on Build GRCh38
rs554765384 Chr.6: 43675280-43675280 on Build GRCh38
rs1012068439 Chr.6: 43675262-43675262 on Build GRCh38
rs773987131 Chr.6: 43675345-43675345 on Build GRCh38
rs745962590 Chr.6: 43675314-43675314 on Build GRCh38
rs35480020 Chr.6: 43675226-43675226 on Build GRCh38
rs1028983643 Chr.6: 43675242-43675242 on Build GRCh38
rs980871175 Chr.6: 43675202-43675203 on Build GRCh38
rs201252905 Chr.6: 43675294-43675294 on Build GRCh38
rs747191839 Chr.6: 43675216-43675216 on Build GRCh38
rs762790208 Chr.6: 43675340-43675340 on Build GRCh38

View Full Product Details