Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 113574204-113574473 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HABP2
Entrez Gene ID
3026
Gene Name
hyaluronan binding protein 2
Gene Aliases
FSAP, HABP, HGFAL, NMTC5, PHBP
UniGene
Hs.422542
Gene Chromosome Location
Chr.10: 113550831-113589602 on Build GRCh38
Transcripts
NM_001177660.2 NM_004132.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5929189 c.154G>A p.E52K Chr.10: 113574336-113574336 on Build GRCh38
4647126 c.194C>T p.P65L Chr.10: 113574376-113574376 on Build GRCh38
383795 c.196G>C p.D66H Chr.10: 113574378-113574378 on Build GRCh38
4569460 c.176T>G p.L59R Chr.10: 113574358-113574358 on Build GRCh38
3769080 c.215A>G p.D72G Chr.10: 113574397-113574397 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749878020 Chr.10: 113574454-113574454 on Build GRCh38
rs746739200 Chr.10: 113574364-113574364 on Build GRCh38
rs757404778 Chr.10: 113574441-113574443 on Build GRCh38
rs763225242 Chr.10: 113574320-113574320 on Build GRCh38
rs759355214 Chr.10: 113574407-113574407 on Build GRCh38
rs914488960 Chr.10: 113574273-113574273 on Build GRCh38
rs756595674 Chr.10: 113574450-113574450 on Build GRCh38
rs1002541034 Chr.10: 113574403-113574403 on Build GRCh38
rs754959601 Chr.10: 113574347-113574347 on Build GRCh38
rs372168368 Chr.10: 113574453-113574453 on Build GRCh38
rs762100939 Chr.10: 113574404-113574404 on Build GRCh38
rs187029293 Chr.10: 113574359-113574359 on Build GRCh38
rs778074411 Chr.10: 113574258-113574258 on Build GRCh38
rs968018428 Chr.10: 113574276-113574276 on Build GRCh38
rs747742981 Chr.10: 113574243-113574243 on Build GRCh38
rs751879626 Chr.10: 113574325-113574325 on Build GRCh38
rs753653917 Chr.10: 113574341-113574341 on Build GRCh38
rs1022952322 Chr.10: 113574433-113574433 on Build GRCh38
rs750872026 Chr.10: 113574289-113574289 on Build GRCh38
rs554833601 Chr.10: 113574301-113574301 on Build GRCh38
rs113225892 Chr.10: 113574330-113574330 on Build GRCh38
rs112910189 Chr.10: 113574281-113574281 on Build GRCh38
rs752239121 Chr.10: 113574351-113574351 on Build GRCh38
rs764200986 Chr.10: 113574266-113574270 on Build GRCh38
rs538121313 Chr.10: 113574251-113574251 on Build GRCh38
rs367719232 Chr.10: 113574421-113574421 on Build GRCh38
rs886046740 Chr.10: 113574340-113574340 on Build GRCh38
rs139478887 Chr.10: 113574389-113574389 on Build GRCh38
rs560894794 Chr.10: 113574409-113574409 on Build GRCh38
rs770453358 Chr.10: 113574384-113574384 on Build GRCh38
rs756954849 Chr.10: 113574426-113574426 on Build GRCh38
rs751896050 Chr.10: 113574281-113574281 on Build GRCh38
rs777386959 Chr.10: 113574362-113574362 on Build GRCh38
rs3740530 Chr.10: 113574365-113574365 on Build GRCh38
rs139657065 Chr.10: 113574321-113574321 on Build GRCh38
rs752684672 Chr.10: 113574414-113574414 on Build GRCh38
rs760090023 Chr.10: 113574295-113574295 on Build GRCh38
rs781369992 Chr.10: 113574431-113574431 on Build GRCh38
rs763138197 Chr.10: 113574253-113574253 on Build GRCh38
rs747308510 Chr.10: 113574263-113574263 on Build GRCh38
rs766265038 Chr.10: 113574332-113574332 on Build GRCh38
rs542838125 Chr.10: 113574405-113574405 on Build GRCh38
rs781628995 Chr.10: 113574368-113574368 on Build GRCh38
rs778401209 Chr.10: 113574239-113574239 on Build GRCh38
rs776996797 Chr.10: 113574280-113574280 on Build GRCh38
rs145969308 Chr.10: 113574382-113574382 on Build GRCh38
rs576302163 Chr.10: 113574425-113574425 on Build GRCh38
rs200071983 Chr.10: 113574385-113574385 on Build GRCh38
rs763595123 Chr.10: 113574423-113574423 on Build GRCh38
rs746211718 Chr.10: 113574435-113574435 on Build GRCh38
rs370803815 Chr.10: 113574309-113574309 on Build GRCh38
rs562968285 Chr.10: 113574443-113574443 on Build GRCh38
rs774432854 Chr.10: 113574393-113574393 on Build GRCh38
rs768915431 Chr.10: 113574390-113574390 on Build GRCh38
rs771380641 Chr.10: 113574270-113574270 on Build GRCh38
rs11575743 Chr.10: 113574237-113574237 on Build GRCh38
rs868670264 Chr.10: 113574375-113574375 on Build GRCh38
rs943468514 Chr.10: 113574305-113574305 on Build GRCh38
rs769702455 Chr.10: 113574308-113574308 on Build GRCh38
rs555658349 Chr.10: 113574235-113574235 on Build GRCh38
rs375680096 Chr.10: 113574331-113574331 on Build GRCh38

View Full Product Details