Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45264847-45265082 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MARK4
Entrez Gene ID
57787
Gene Name
microtubule affinity regulating kinase 4
Gene Aliases
MARK4L, MARK4S, MARKL1, MARKL1L, PAR-1D
UniGene
Hs.34314
Gene Chromosome Location
Chr.19: 45251258-45305283 on Build GRCh38
Transcripts
NM_001199867.1 NM_031417.3 XM_006723307.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3103903 c.446C>T p.S149L Chr.19: 45264864-45264864 on Build GRCh38
3103906 c.447G>A p.S149S Chr.19: 45264865-45264865 on Build GRCh38
3103905 c.447G>A p.S149S Chr.19: 45264865-45264865 on Build GRCh38
5564632 c.488G>A p.R163Q Chr.19: 45264906-45264906 on Build GRCh38
3103900 c.441C>T p.L147L Chr.19: 45264859-45264859 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774897035 Chr.19: 45264874-45264874 on Build GRCh38
rs966167593 Chr.19: 45265035-45265035 on Build GRCh38
rs764261390 Chr.19: 45264927-45264927 on Build GRCh38
rs759837890 Chr.19: 45264877-45264877 on Build GRCh38
rs760927301 Chr.19: 45264919-45264919 on Build GRCh38
rs781065294 Chr.19: 45264961-45264961 on Build GRCh38
rs775536202 Chr.19: 45264917-45264917 on Build GRCh38
rs373067738 Chr.19: 45264872-45264872 on Build GRCh38
rs1044535314 Chr.19: 45264916-45264916 on Build GRCh38
rs565736296 Chr.19: 45264894-45264894 on Build GRCh38
rs967720850 Chr.19: 45265014-45265014 on Build GRCh38
rs1013015449 Chr.19: 45264913-45264913 on Build GRCh38
rs1037111543 Chr.19: 45264978-45264978 on Build GRCh38
rs866598034 Chr.19: 45264999-45264999 on Build GRCh38
rs757465821 Chr.19: 45264928-45264928 on Build GRCh38
rs976666797 Chr.19: 45265046-45265046 on Build GRCh38
rs754602902 Chr.19: 45264962-45264962 on Build GRCh38
rs985839246 Chr.19: 45265015-45265015 on Build GRCh38
rs995489016 Chr.19: 45264996-45264996 on Build GRCh38
rs866208477 Chr.19: 45264873-45264873 on Build GRCh38
rs180708356 Chr.19: 45264998-45264998 on Build GRCh38
rs897159543 Chr.19: 45264979-45264979 on Build GRCh38
rs766356561 Chr.19: 45264939-45264939 on Build GRCh38
rs757401170 Chr.19: 45264938-45264939 on Build GRCh38
rs890059107 Chr.19: 45265004-45265004 on Build GRCh38
rs1009196023 Chr.19: 45264971-45264971 on Build GRCh38
rs754685856 Chr.19: 45264952-45264952 on Build GRCh38
rs751544600 Chr.19: 45264951-45264951 on Build GRCh38
rs948529215 Chr.19: 45264907-45264907 on Build GRCh38
rs910324074 Chr.19: 45265019-45265019 on Build GRCh38
rs373844257 Chr.19: 45264964-45264964 on Build GRCh38

View Full Product Details