Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1984252-1984497 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GFER
Entrez Gene ID
2671
Gene Name
growth factor, augmenter of liver regeneration
Gene Aliases
ALR, ERV1, HERV1, HPO, HPO1, HPO2, HSS
UniGene
Hs.27184
Gene Chromosome Location
Chr.16: 1984149-1987749 on Build GRCh38
Transcripts
NM_005262.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6398493 c.88C>A p.L30M Chr.16: 1984306-1984306 on Build GRCh38
6500099 c.50T>A p.F17Y Chr.16: 1984268-1984268 on Build GRCh38
397508 c.215G>T p.R72L Chr.16: 1984433-1984433 on Build GRCh38
5363495 c.218C>T p.A73V Chr.16: 1984436-1984436 on Build GRCh38
7320582 c.176C>T p.S59F Chr.16: 1984394-1984394 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs535377905 Chr.16: 1984275-1984275 on Build GRCh38
rs1042664976 Chr.16: 1984276-1984276 on Build GRCh38
rs863224028 Chr.16: 1984417-1984417 on Build GRCh38
rs773199934 Chr.16: 1984280-1984280 on Build GRCh38
rs1028643548 Chr.16: 1984421-1984421 on Build GRCh38
rs955632773 Chr.16: 1984401-1984401 on Build GRCh38
rs759460274 Chr.16: 1984371-1984371 on Build GRCh38
rs780490309 Chr.16: 1984427-1984427 on Build GRCh38
rs910026091 Chr.16: 1984305-1984305 on Build GRCh38
rs758150888 Chr.16: 1984386-1984386 on Build GRCh38
rs1003355020 Chr.16: 1984298-1984298 on Build GRCh38
rs893122483 Chr.16: 1984273-1984273 on Build GRCh38
rs748329975 Chr.16: 1984442-1984442 on Build GRCh38
rs1011987202 Chr.16: 1984341-1984341 on Build GRCh38
rs950089553 Chr.16: 1984345-1984345 on Build GRCh38
rs770900182 Chr.16: 1984477-1984477 on Build GRCh38
rs901752142 Chr.16: 1984411-1984411 on Build GRCh38
rs766280079 Chr.16: 1984320-1984320 on Build GRCh38
rs972973281 Chr.16: 1984316-1984316 on Build GRCh38
rs201675904 Chr.16: 1984462-1984462 on Build GRCh38
rs936063405 Chr.16: 1984356-1984356 on Build GRCh38
rs863224027 Chr.16: 1984463-1984463 on Build GRCh38
rs749652504 Chr.16: 1984432-1984432 on Build GRCh38
rs972426940 Chr.16: 1984361-1984361 on Build GRCh38
rs1010571528 Chr.16: 1984394-1984394 on Build GRCh38
rs760615529 Chr.16: 1984281-1984281 on Build GRCh38
rs752327413 Chr.16: 1984377-1984377 on Build GRCh38
rs773323504 Chr.16: 1984454-1984454 on Build GRCh38
rs763610770 Chr.16: 1984404-1984404 on Build GRCh38
rs918808957 Chr.16: 1984372-1984372 on Build GRCh38
rs1034865237 Chr.16: 1984308-1984308 on Build GRCh38
rs765134997 Chr.16: 1984375-1984375 on Build GRCh38
rs902431596 Chr.16: 1984293-1984293 on Build GRCh38
rs375792737 Chr.16: 1984407-1984407 on Build GRCh38
rs891592950 Chr.16: 1984364-1984364 on Build GRCh38
rs755279097 Chr.16: 1984434-1984434 on Build GRCh38
rs753639816 Chr.16: 1984334-1984334 on Build GRCh38
rs772307688 Chr.16: 1984443-1984443 on Build GRCh38
rs1016725284 Chr.16: 1984347-1984347 on Build GRCh38
rs779254345 Chr.16: 1984435-1984435 on Build GRCh38
rs112534956 Chr.16: 1984410-1984410 on Build GRCh38
rs569037883 Chr.16: 1984423-1984423 on Build GRCh38
rs866480610 Chr.16: 1984449-1984449 on Build GRCh38
rs962686909 Chr.16: 1984351-1984351 on Build GRCh38

View Full Product Details