Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 57262040-57262299 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MSI2
Entrez Gene ID
124540
Gene Name
musashi RNA binding protein 2
Gene Aliases
MSI2H
UniGene
Hs.658922
Gene Chromosome Location
Chr.17: 57255851-57684689 on Build GRCh38
Transcripts
NM_001322250.1 NM_001322251.1 NM_138962.3 NM_170721.1 XM_005257014.3 XM_005257015.3 XM_011524286.1 XM_017024147.1 XM_017024148.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1286284 c.286C>T p.R96* Chr.17: 57262178-57262178 on Build GRCh38
5905319 c.271-4C>T p.? Chr.17: 57262147-57262147 on Build GRCh38
1130223 c.303G>A p.A101A Chr.17: 57262183-57262183 on Build GRCh38
6998848 c.283C>G p.R95G Chr.17: 57262175-57262175 on Build GRCh38
6998847 c.295C>G p.R99G Chr.17: 57262175-57262175 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749230653 Chr.17: 57262137-57262138 on Build GRCh38
rs368855513 Chr.17: 57262244-57262244 on Build GRCh38
rs373775343 Chr.17: 57262094-57262094 on Build GRCh38
rs746335310 Chr.17: 57262235-57262235 on Build GRCh38
rs771413443 Chr.17: 57262118-57262118 on Build GRCh38
rs777873590 Chr.17: 57262211-57262211 on Build GRCh38
rs772621490 Chr.17: 57262236-57262236 on Build GRCh38
rs774744753 Chr.17: 57262120-57262120 on Build GRCh38
rs577350868 Chr.17: 57262252-57262252 on Build GRCh38
rs775827378 Chr.17: 57262243-57262243 on Build GRCh38
rs749342681 Chr.17: 57262227-57262227 on Build GRCh38
rs374657165 Chr.17: 57262115-57262115 on Build GRCh38
rs766301726 Chr.17: 57262203-57262203 on Build GRCh38
rs746201359 Chr.17: 57262123-57262123 on Build GRCh38
rs1031184752 Chr.17: 57262127-57262127 on Build GRCh38
rs781442048 Chr.17: 57262261-57262261 on Build GRCh38
rs979248656 Chr.17: 57262119-57262119 on Build GRCh38
rs948348306 Chr.17: 57262185-57262185 on Build GRCh38
rs775878253 Chr.17: 57262130-57262130 on Build GRCh38
rs758633174 Chr.17: 57262116-57262116 on Build GRCh38
rs780206273 Chr.17: 57262126-57262126 on Build GRCh38
rs200593685 Chr.17: 57262137-57262137 on Build GRCh38
rs757310411 Chr.17: 57262230-57262230 on Build GRCh38
rs140648416 Chr.17: 57262208-57262208 on Build GRCh38
rs574167789 Chr.17: 57262183-57262183 on Build GRCh38
rs764936146 Chr.17: 57262138-57262138 on Build GRCh38
rs1036509815 Chr.17: 57262276-57262276 on Build GRCh38
rs947718601 Chr.17: 57262269-57262269 on Build GRCh38
rs555639930 Chr.17: 57262165-57262165 on Build GRCh38
rs756342396 Chr.17: 57262210-57262210 on Build GRCh38
rs778001370 Chr.17: 57262138-57262138 on Build GRCh38
rs779087184 Chr.17: 57262233-57262233 on Build GRCh38
rs772710603 Chr.17: 57262182-57262182 on Build GRCh38
rs34687737 Chr.17: 57262113-57262113 on Build GRCh38
rs747256635 Chr.17: 57262147-57262147 on Build GRCh38
rs751423453 Chr.17: 57262205-57262205 on Build GRCh38
rs955384023 Chr.17: 57262070-57262076 on Build GRCh38
rs200133663 Chr.17: 57262209-57262209 on Build GRCh38
rs755014494 Chr.17: 57262207-57262207 on Build GRCh38

View Full Product Details