Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 23569621-23569858 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JPH4
Entrez Gene ID
84502
Gene Name
junctophilin 4
Gene Aliases
JP4, JPHL1
UniGene
Hs.597841
Gene Chromosome Location
Chr.14: 23568035-23578800 on Build GRCh38
Transcripts
NM_001146028.1 NM_032452.2

SNP Detail

SNP ID Chromosome Location
rs895630984 Chr.14: 23569702-23569702 on Build GRCh38
rs374032273 Chr.14: 23569832-23569832 on Build GRCh38
rs776314618 Chr.14: 23569741-23569741 on Build GRCh38
rs374439117 Chr.14: 23569740-23569740 on Build GRCh38
rs766269080 Chr.14: 23569728-23569729 on Build GRCh38
rs989046613 Chr.14: 23569735-23569735 on Build GRCh38
rs1000255769 Chr.14: 23569757-23569757 on Build GRCh38
rs139748760 Chr.14: 23569730-23569730 on Build GRCh38
rs760356106 Chr.14: 23569650-23569650 on Build GRCh38
rs865865801 Chr.14: 23569711-23569711 on Build GRCh38
rs74587188 Chr.14: 23569753-23569753 on Build GRCh38
rs1036956580 Chr.14: 23569688-23569688 on Build GRCh38
rs766161223 Chr.14: 23569777-23569777 on Build GRCh38
rs572663157 Chr.14: 23569800-23569800 on Build GRCh38
rs144807486 Chr.14: 23569679-23569679 on Build GRCh38
rs371235987 Chr.14: 23569676-23569676 on Build GRCh38
rs949933230 Chr.14: 23569779-23569779 on Build GRCh38

View Full Product Details