Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 47410145-47410412 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MSH2
Entrez Gene ID
4436
Gene Name
mutS homolog 2
Gene Aliases
COCA1, FCC1, HNPCC, HNPCC1, LCFS2
UniGene
Hs.597656
Gene Chromosome Location
Chr.2: 47403067-47634501 on Build GRCh38
Transcripts
NM_000251.2 NM_001258281.1 XM_005264332.3 XM_011532867.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6951777 c.478C>T p.Q160* Chr.2: 47410205-47410205 on Build GRCh38
6975371 c.460G>T p.A154S Chr.2: 47410187-47410187 on Build GRCh38
1684714 c.505A>G p.I169V Chr.2: 47410232-47410232 on Build GRCh38
6421877 c.620C>A p.A207D Chr.2: 47410347-47410347 on Build GRCh38
6950715 c.568C>T p.L190F Chr.2: 47410295-47410295 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs730881778 Chr.2: 47410308-47410308 on Build GRCh38
rs587779973 Chr.2: 47410334-47410334 on Build GRCh38
rs786204319 Chr.2: 47410202-47410203 on Build GRCh38
rs63750913 Chr.2: 47410346-47410346 on Build GRCh38
rs528114416 Chr.2: 47410270-47410270 on Build GRCh38
rs764573221 Chr.2: 47410297-47410297 on Build GRCh38
rs151129360 Chr.2: 47410284-47410284 on Build GRCh38
rs140217708 Chr.2: 47410380-47410380 on Build GRCh38
rs63751326 Chr.2: 47410304-47410304 on Build GRCh38
rs587779163 Chr.2: 47410220-47410220 on Build GRCh38
rs770787472 Chr.2: 47410338-47410338 on Build GRCh38
rs149511545 Chr.2: 47410208-47410208 on Build GRCh38
rs63751444 Chr.2: 47410287-47410287 on Build GRCh38
rs63750070 Chr.2: 47410245-47410245 on Build GRCh38
rs587780689 Chr.2: 47410358-47410358 on Build GRCh38
rs746013810 Chr.2: 47410352-47410352 on Build GRCh38
rs63749984 Chr.2: 47410340-47410340 on Build GRCh38
rs63751291 Chr.2: 47410251-47410251 on Build GRCh38
rs759603999 Chr.2: 47410286-47410286 on Build GRCh38
rs63751136 Chr.2: 47410323-47410323 on Build GRCh38
rs63750624 Chr.2: 47410211-47410211 on Build GRCh38
rs786203795 Chr.2: 47410257-47410257 on Build GRCh38
rs63750574 Chr.2: 47410337-47410337 on Build GRCh38
rs63751449 Chr.2: 47410181-47410181 on Build GRCh38
rs929758799 Chr.2: 47410172-47410172 on Build GRCh38
rs902336078 Chr.2: 47410263-47410263 on Build GRCh38
rs876659506 Chr.2: 47410318-47410318 on Build GRCh38
rs63750705 Chr.2: 47410250-47410291 on Build GRCh38
rs63751622 Chr.2: 47410365-47410366 on Build GRCh38
rs63750995 Chr.2: 47410343-47410344 on Build GRCh38
rs63750551 Chr.2: 47410257-47410258 on Build GRCh38
rs63750121 Chr.2: 47410350-47410350 on Build GRCh38
rs587779165 Chr.2: 47410298-47410300 on Build GRCh38
rs876658623 Chr.2: 47410344-47410344 on Build GRCh38
rs63750682 Chr.2: 47410314-47410314 on Build GRCh38
rs63750126 Chr.2: 47410209-47410209 on Build GRCh38
rs878853819 Chr.2: 47410281-47410281 on Build GRCh38
rs63750773 Chr.2: 47410212-47410212 on Build GRCh38
rs786204082 Chr.2: 47410218-47410218 on Build GRCh38
rs63750382 Chr.2: 47410256-47410256 on Build GRCh38
rs63750255 Chr.2: 47410226-47410226 on Build GRCh38
rs141021599 Chr.2: 47410293-47410293 on Build GRCh38
rs267607689 Chr.2: 47410373-47410373 on Build GRCh38
rs587779168 Chr.2: 47410375-47410375 on Build GRCh38
rs754478179 Chr.2: 47410314-47410314 on Build GRCh38
rs63751110 Chr.2: 47410322-47410322 on Build GRCh38
rs63750600 Chr.2: 47410333-47410333 on Build GRCh38
rs587779164 Chr.2: 47410255-47410256 on Build GRCh38
rs751250018 Chr.2: 47410366-47410366 on Build GRCh38
rs63750902 Chr.2: 47410239-47410239 on Build GRCh38
rs763459034 Chr.2: 47410295-47410295 on Build GRCh38
rs63750327 Chr.2: 47410320-47410320 on Build GRCh38
rs63750080 Chr.2: 47410291-47410292 on Build GRCh38
rs748762580 Chr.2: 47410234-47410234 on Build GRCh38
rs781178004 Chr.2: 47410347-47410347 on Build GRCh38
rs786202238 Chr.2: 47410279-47410279 on Build GRCh38
rs63751426 Chr.2: 47410205-47410205 on Build GRCh38
rs63750786 Chr.2: 47410319-47410320 on Build GRCh38
rs63750037 Chr.2: 47410274-47410274 on Build GRCh38
rs587779166 Chr.2: 47410319-47410319 on Build GRCh38
rs864622381 Chr.2: 47410303-47410303 on Build GRCh38
rs730881770 Chr.2: 47410271-47410271 on Build GRCh38
rs63749949 Chr.2: 47410222-47410222 on Build GRCh38
rs767972932 Chr.2: 47410388-47410388 on Build GRCh38
rs778573140 Chr.2: 47410316-47410316 on Build GRCh38
rs769971586 Chr.2: 47410357-47410357 on Build GRCh38
rs763298811 Chr.2: 47410368-47410368 on Build GRCh38
rs765489269 Chr.2: 47410197-47410197 on Build GRCh38
rs766497093 Chr.2: 47410283-47410283 on Build GRCh38
rs587779162 Chr.2: 47410173-47410173 on Build GRCh38
rs587782804 Chr.2: 47410313-47410313 on Build GRCh38
rs876658188 Chr.2: 47410191-47410191 on Build GRCh38
rs757733033 Chr.2: 47410228-47410228 on Build GRCh38
rs375202935 Chr.2: 47410210-47410210 on Build GRCh38
rs369685768 Chr.2: 47410321-47410321 on Build GRCh38
rs147346837 Chr.2: 47410248-47410248 on Build GRCh38
rs63750933 Chr.2: 47410240-47410240 on Build GRCh38
rs63751065 Chr.2: 47410186-47410186 on Build GRCh38
rs587779167 Chr.2: 47410326-47410326 on Build GRCh38
rs63751695 Chr.2: 47410369-47410372 on Build GRCh38
rs768006988 Chr.2: 47410301-47410301 on Build GRCh38
rs786203142 Chr.2: 47410174-47410174 on Build GRCh38
rs63750716 Chr.2: 47410232-47410232 on Build GRCh38
rs63750069 Chr.2: 47410245-47410245 on Build GRCh38
rs954909694 Chr.2: 47410288-47410288 on Build GRCh38
rs1800151 Chr.2: 47410300-47410300 on Build GRCh38
rs63750088 Chr.2: 47410288-47410296 on Build GRCh38
rs267607928 Chr.2: 47410278-47410278 on Build GRCh38
rs63750582 Chr.2: 47410217-47410217 on Build GRCh38
rs786202651 Chr.2: 47410351-47410351 on Build GRCh38
rs63751013 Chr.2: 47410233-47410236 on Build GRCh38
rs766349734 Chr.2: 47410185-47410185 on Build GRCh38
rs876658996 Chr.2: 47410374-47410374 on Build GRCh38
rs759712763 Chr.2: 47410187-47410187 on Build GRCh38
rs63751226 Chr.2: 47410199-47410199 on Build GRCh38
rs63751067 Chr.2: 47410244-47410255 on Build GRCh38
rs63750843 Chr.2: 47410235-47410235 on Build GRCh38
rs63750214 Chr.2: 47410215-47410215 on Build GRCh38
rs768931909 Chr.2: 47410369-47410369 on Build GRCh38
rs63750821 Chr.2: 47410292-47410292 on Build GRCh38
rs786202921 Chr.2: 47410203-47410203 on Build GRCh38
rs63751274 Chr.2: 47410370-47410370 on Build GRCh38
rs771827041 Chr.2: 47410252-47410252 on Build GRCh38
rs762825105 Chr.2: 47410196-47410198 on Build GRCh38
rs267607927 Chr.2: 47410296-47410297 on Build GRCh38
rs61756463 Chr.2: 47410198-47410198 on Build GRCh38

View Full Product Details