Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 142864190-142864436 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHB6
Entrez Gene ID
2051
Gene Name
EPH receptor B6
Gene Aliases
HEP
UniGene
Hs.380089
Gene Chromosome Location
Chr.7: 142854640-142871093 on Build GRCh38
Transcripts
NM_001280794.2 NM_001280795.2 NM_004445.5 NR_104001.2 XM_006715881.2 XM_011515879.1 XM_011515880.1 XM_011515881.2 XM_011515882.2

SNP Detail

SNP ID Chromosome Location
rs763236627 Chr.7: 142864337-142864337 on Build GRCh38
rs777151089 Chr.7: 142864272-142864272 on Build GRCh38
rs781406143 Chr.7: 142864349-142864350 on Build GRCh38
rs755315060 Chr.7: 142864211-142864211 on Build GRCh38
rs149090003 Chr.7: 142864399-142864399 on Build GRCh38
rs779876904 Chr.7: 142864306-142864306 on Build GRCh38
rs192960264 Chr.7: 142864300-142864300 on Build GRCh38
rs777875083 Chr.7: 142864219-142864219 on Build GRCh38
rs778542770 Chr.7: 142864246-142864246 on Build GRCh38
rs151277713 Chr.7: 142864229-142864229 on Build GRCh38
rs145358081 Chr.7: 142864252-142864252 on Build GRCh38
rs141491735 Chr.7: 142864336-142864336 on Build GRCh38
rs762655099 Chr.7: 142864330-142864330 on Build GRCh38
rs753306588 Chr.7: 142864297-142864302 on Build GRCh38
rs752558281 Chr.7: 142864371-142864371 on Build GRCh38
rs778827216 Chr.7: 142864315-142864315 on Build GRCh38
rs750457941 Chr.7: 142864356-142864356 on Build GRCh38
rs764396514 Chr.7: 142864300-142864301 on Build GRCh38
rs267601348 Chr.7: 142864303-142864303 on Build GRCh38
rs536789493 Chr.7: 142864301-142864301 on Build GRCh38
rs537204866 Chr.7: 142864245-142864245 on Build GRCh38
rs775969564 Chr.7: 142864332-142864332 on Build GRCh38
rs765672490 Chr.7: 142864283-142864283 on Build GRCh38
rs56151513 Chr.7: 142864311-142864311 on Build GRCh38
rs749048034 Chr.7: 142864231-142864231 on Build GRCh38
rs942674128 Chr.7: 142864297-142864308 on Build GRCh38
rs751901904 Chr.7: 142864307-142864308 on Build GRCh38
rs370835347 Chr.7: 142864230-142864230 on Build GRCh38
rs143416563 Chr.7: 142864325-142864325 on Build GRCh38
rs770338059 Chr.7: 142864403-142864403 on Build GRCh38
rs868369267 Chr.7: 142864291-142864291 on Build GRCh38
rs571330909 Chr.7: 142864334-142864334 on Build GRCh38
rs375814622 Chr.7: 142864388-142864388 on Build GRCh38
rs8177142 Chr.7: 142864298-142864298 on Build GRCh38
rs8177141 Chr.7: 142864253-142864253 on Build GRCh38
rs764828501 Chr.7: 142864297-142864299 on Build GRCh38
rs78315767 Chr.7: 142864322-142864322 on Build GRCh38
rs757433149 Chr.7: 142864316-142864317 on Build GRCh38
rs759845995 Chr.7: 142864277-142864277 on Build GRCh38
rs139237333 Chr.7: 142864251-142864251 on Build GRCh38
rs868655664 Chr.7: 142864307-142864307 on Build GRCh38
rs746542529 Chr.7: 142864328-142864328 on Build GRCh38
rs751729707 Chr.7: 142864349-142864349 on Build GRCh38
rs763215082 Chr.7: 142864297-142864297 on Build GRCh38
rs146506957 Chr.7: 142864237-142864237 on Build GRCh38
rs146978351 Chr.7: 142864355-142864355 on Build GRCh38
rs369872682 Chr.7: 142864398-142864398 on Build GRCh38
rs375498604 Chr.7: 142864258-142864258 on Build GRCh38
rs770184288 Chr.7: 142864335-142864335 on Build GRCh38
rs764313153 Chr.7: 142864347-142864347 on Build GRCh38
rs747892419 Chr.7: 142864321-142864321 on Build GRCh38
rs753854523 Chr.7: 142864310-142864310 on Build GRCh38
rs142367849 Chr.7: 142864214-142864214 on Build GRCh38
rs555319297 Chr.7: 142864417-142864417 on Build GRCh38
rs61732974 Chr.7: 142864215-142864215 on Build GRCh38
rs774101812 Chr.7: 142864368-142864368 on Build GRCh38
rs148373849 Chr.7: 142864367-142864367 on Build GRCh38
rs765403799 Chr.7: 142864212-142864212 on Build GRCh38
rs771581358 Chr.7: 142864266-142864266 on Build GRCh38
rs770528745 Chr.7: 142864329-142864329 on Build GRCh38
rs958321807 Chr.7: 142864213-142864213 on Build GRCh38
rs763526105 Chr.7: 142864284-142864284 on Build GRCh38
rs746695848 Chr.7: 142864393-142864393 on Build GRCh38
rs200894619 Chr.7: 142864389-142864389 on Build GRCh38
rs756835314 Chr.7: 142864396-142864396 on Build GRCh38

View Full Product Details