Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12875771-12876044 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNASE2
Entrez Gene ID
1777
Gene Name
deoxyribonuclease II, lysosomal
Gene Aliases
DNASE2A, DNL, DNL2
UniGene
Hs.118243
Gene Chromosome Location
Chr.19: 12875211-12881521 on Build GRCh38
Transcripts
NM_001375.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6419318 c.1050G>T p.M350I Chr.19: 12876023-12876023 on Build GRCh38
1204235 c.1040G>A p.C347Y Chr.19: 12876033-12876033 on Build GRCh38
2816840 c.1038C>A p.P346P Chr.19: 12876035-12876035 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs759870950 Chr.19: 12875983-12875983 on Build GRCh38
rs928679043 Chr.19: 12875925-12875925 on Build GRCh38
rs991536297 Chr.19: 12875958-12875958 on Build GRCh38
rs79595281 Chr.19: 12875883-12875883 on Build GRCh38
rs933463118 Chr.19: 12875971-12875971 on Build GRCh38
rs911860695 Chr.19: 12875897-12875897 on Build GRCh38
rs576215922 Chr.19: 12875909-12875909 on Build GRCh38
rs775947025 Chr.19: 12875961-12875961 on Build GRCh38
rs914552111 Chr.19: 12875960-12875960 on Build GRCh38
rs953620543 Chr.19: 12875867-12875867 on Build GRCh38
rs554384572 Chr.19: 12875994-12875994 on Build GRCh38
rs752955913 Chr.19: 12876009-12876009 on Build GRCh38
rs978909242 Chr.19: 12875835-12875835 on Build GRCh38
rs771385627 Chr.19: 12875970-12875970 on Build GRCh38
rs1016329104 Chr.19: 12875793-12875793 on Build GRCh38
rs1002518341 Chr.19: 12876013-12876013 on Build GRCh38
rs144569909 Chr.19: 12876014-12876014 on Build GRCh38
rs776917620 Chr.19: 12875979-12875979 on Build GRCh38
rs919347845 Chr.19: 12875937-12875937 on Build GRCh38
rs970512070 Chr.19: 12875889-12875889 on Build GRCh38
rs944741902 Chr.19: 12875902-12875902 on Build GRCh38
rs963359037 Chr.19: 12875802-12875802 on Build GRCh38
rs759318810 Chr.19: 12875865-12875865 on Build GRCh38
rs937389483 Chr.19: 12875936-12875936 on Build GRCh38
rs192025167 Chr.19: 12875916-12875916 on Build GRCh38
rs538894338 Chr.19: 12875855-12875855 on Build GRCh38
rs768199297 Chr.19: 12875947-12875947 on Build GRCh38
rs967493166 Chr.19: 12875826-12875826 on Build GRCh38
rs368029639 Chr.19: 12875976-12875976 on Build GRCh38
rs200124589 Chr.19: 12875963-12875963 on Build GRCh38

View Full Product Details