Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 92091779-92092032 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPEB3
Entrez Gene ID
22849
Gene Name
cytoplasmic polyadenylation element binding protein 3
Gene Aliases
-
UniGene
Hs.131683
Gene Chromosome Location
Chr.10: 92046692-92291118 on Build GRCh38
Transcripts
NM_001178137.1 NM_014912.4 XM_005269630.4 XM_006717714.3 XM_006717715.3 XM_011539514.2 XM_011539515.2 XM_011539517.2 XM_011539518.2 XM_011539519.2 XM_011539520.2 XM_017015927.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4529807 c.1643G>A p.R548K Chr.10: 92091874-92091874 on Build GRCh38
7329769 c.1558G>A p.D520N Chr.10: 92091917-92091917 on Build GRCh38
6634544 c.1558G>T p.D520Y Chr.10: 92091917-92091917 on Build GRCh38
6634552 c.1580T>A p.I527N Chr.10: 92091937-92091937 on Build GRCh38
4182218 c.1568T>G p.F523C Chr.10: 92091907-92091907 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759894214 Chr.10: 92091978-92091979 on Build GRCh38
rs758449954 Chr.10: 92091987-92091987 on Build GRCh38
rs760097953 Chr.10: 92091819-92091819 on Build GRCh38
rs577416184 Chr.10: 92091849-92091849 on Build GRCh38
rs767977151 Chr.10: 92091836-92091836 on Build GRCh38
rs971951296 Chr.10: 92091845-92091845 on Build GRCh38
rs774855993 Chr.10: 92091816-92091816 on Build GRCh38
rs780119787 Chr.10: 92091992-92091992 on Build GRCh38
rs993640710 Chr.10: 92091916-92091916 on Build GRCh38
rs775592518 Chr.10: 92091841-92091841 on Build GRCh38
rs369153928 Chr.10: 92091930-92091930 on Build GRCh38
rs35476882 Chr.10: 92092004-92092005 on Build GRCh38
rs750240026 Chr.10: 92091958-92091958 on Build GRCh38
rs376994406 Chr.10: 92091986-92091986 on Build GRCh38
rs764406144 Chr.10: 92091864-92091864 on Build GRCh38
rs374710766 Chr.10: 92091808-92091808 on Build GRCh38
rs1229578 Chr.10: 92091957-92091957 on Build GRCh38

View Full Product Details