Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 76449492-76449763 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNTNAP4
Entrez Gene ID
85445
Gene Name
contactin associated protein like 4
Gene Aliases
CASPR4
UniGene
Hs.461389
Gene Chromosome Location
Chr.16: 76277217-76563610 on Build GRCh38
Transcripts
NM_001322178.1 NM_001322179.1 NM_001322180.1 NM_001322181.1 NM_001322187.1 NM_001322188.1 NM_001322189.1 NM_001322190.1 NM_001322191.1 NM_033401.4 NM_138994.4 NR_136210.1 NR_136213.1 NR_136216.1 XM_011523403.2 XM_017023805.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5722486 c.881G>A p.G294E Chr.16: 76449743-76449743 on Build GRCh38
3512375 c.897C>T p.F299F Chr.16: 76449759-76449759 on Build GRCh38
2691823 c.868A>T p.I290F Chr.16: 76449730-76449730 on Build GRCh38
4432496 c.878C>A p.P293H Chr.16: 76449740-76449740 on Build GRCh38
7318024 c.965C>T p.S322L Chr.16: 76449755-76449755 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs74025004 Chr.16: 76449584-76449584 on Build GRCh38
rs1051370381 Chr.16: 76449651-76449651 on Build GRCh38
rs760416068 Chr.16: 76449606-76449606 on Build GRCh38
rs1015016465 Chr.16: 76449655-76449655 on Build GRCh38
rs370684680 Chr.16: 76449692-76449692 on Build GRCh38
rs879348816 Chr.16: 76449618-76449618 on Build GRCh38
rs559425904 Chr.16: 76449637-76449637 on Build GRCh38
rs376251104 Chr.16: 76449706-76449706 on Build GRCh38
rs987677966 Chr.16: 76449705-76449705 on Build GRCh38
rs890079276 Chr.16: 76449652-76449652 on Build GRCh38
rs780076343 Chr.16: 76449727-76449727 on Build GRCh38
rs948765167 Chr.16: 76449532-76449532 on Build GRCh38
rs545730614 Chr.16: 76449683-76449683 on Build GRCh38
rs936153941 Chr.16: 76449594-76449594 on Build GRCh38
rs138261248 Chr.16: 76449593-76449593 on Build GRCh38
rs542661291 Chr.16: 76449619-76449619 on Build GRCh38
rs768605721 Chr.16: 76449663-76449663 on Build GRCh38
rs374023701 Chr.16: 76449591-76449591 on Build GRCh38
rs963137057 Chr.16: 76449664-76449664 on Build GRCh38
rs760143846 Chr.16: 76449666-76449666 on Build GRCh38
rs572854344 Chr.16: 76449681-76449681 on Build GRCh38
rs917208569 Chr.16: 76449524-76449524 on Build GRCh38
rs1007188661 Chr.16: 76449658-76449658 on Build GRCh38
rs764357310 Chr.16: 76449688-76449688 on Build GRCh38
rs1053305240 Chr.16: 76449616-76449616 on Build GRCh38
rs983374714 Chr.16: 76449640-76449640 on Build GRCh38
rs371694462 Chr.16: 76449736-76449736 on Build GRCh38
rs149277788 Chr.16: 76449597-76449597 on Build GRCh38
rs959970878 Chr.16: 76449620-76449620 on Build GRCh38
rs905712007 Chr.16: 76449602-76449602 on Build GRCh38
rs1026594436 Chr.16: 76449684-76449684 on Build GRCh38
rs557930683 Chr.16: 76449686-76449686 on Build GRCh38
rs763443174 Chr.16: 76449685-76449685 on Build GRCh38
rs768548223 Chr.16: 76449674-76449677 on Build GRCh38
rs1001354791 Chr.16: 76449603-76449603 on Build GRCh38
rs375068105 Chr.16: 76449700-76449700 on Build GRCh38
rs756839864 Chr.16: 76449697-76449697 on Build GRCh38
rs992828674 Chr.16: 76449514-76449514 on Build GRCh38
rs370295203 Chr.16: 76449709-76449709 on Build GRCh38
rs755597174 Chr.16: 76449714-76449714 on Build GRCh38
rs998816907 Chr.16: 76449644-76449644 on Build GRCh38
rs970009969 Chr.16: 76449739-76449739 on Build GRCh38
rs538421692 Chr.16: 76449600-76449601 on Build GRCh38

View Full Product Details