Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 92450604-92450853 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GATAD1
Entrez Gene ID
57798
Gene Name
GATA zinc finger domain containing 1
Gene Aliases
CMD2B, ODAG, RG083M05.2
UniGene
Hs.21145
Gene Chromosome Location
Chr.7: 92447448-92494631 on Build GRCh38
Transcripts
NM_021167.4 NR_052016.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4686492 c.407T>A p.I136K Chr.7: 92450732-92450732 on Build GRCh38
3703308 c.376-8T>G p.? Chr.7: 92450693-92450693 on Build GRCh38
1452832 c.378C>T p.P126P Chr.7: 92450703-92450703 on Build GRCh38
6111430 c.381C>G p.I127M Chr.7: 92450706-92450706 on Build GRCh38
3642258 c.411C>T p.I137I Chr.7: 92450736-92450736 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769037886 Chr.7: 92450636-92450636 on Build GRCh38
rs183288154 Chr.7: 92450653-92450653 on Build GRCh38
rs749450092 Chr.7: 92450772-92450772 on Build GRCh38
rs775799869 Chr.7: 92450809-92450809 on Build GRCh38
rs1014941169 Chr.7: 92450703-92450703 on Build GRCh38
rs774855382 Chr.7: 92450651-92450651 on Build GRCh38
rs142445224 Chr.7: 92450823-92450823 on Build GRCh38
rs369519708 Chr.7: 92450798-92450798 on Build GRCh38
rs375155246 Chr.7: 92450816-92450816 on Build GRCh38
rs756899166 Chr.7: 92450669-92450669 on Build GRCh38
rs889121199 Chr.7: 92450676-92450676 on Build GRCh38
rs774767272 Chr.7: 92450780-92450780 on Build GRCh38
rs200584566 Chr.7: 92450756-92450756 on Build GRCh38
rs771250518 Chr.7: 92450776-92450776 on Build GRCh38
rs753400453 Chr.7: 92450668-92450668 on Build GRCh38
rs753276680 Chr.7: 92450658-92450658 on Build GRCh38
rs779817755 Chr.7: 92450749-92450749 on Build GRCh38
rs904655991 Chr.7: 92450646-92450646 on Build GRCh38
rs958141257 Chr.7: 92450704-92450704 on Build GRCh38
rs772487915 Chr.7: 92450807-92450807 on Build GRCh38
rs759773637 Chr.7: 92450800-92450800 on Build GRCh38
rs200665705 Chr.7: 92450731-92450731 on Build GRCh38
rs377121457 Chr.7: 92450765-92450765 on Build GRCh38
rs760033926 Chr.7: 92450659-92450659 on Build GRCh38
rs876657464 Chr.7: 92450775-92450776 on Build GRCh38
rs191104636 Chr.7: 92450645-92450645 on Build GRCh38
rs1021060263 Chr.7: 92450777-92450777 on Build GRCh38
rs777986941 Chr.7: 92450770-92450770 on Build GRCh38
rs372313754 Chr.7: 92450663-92450663 on Build GRCh38
rs876657814 Chr.7: 92450688-92450689 on Build GRCh38
rs376691322 Chr.7: 92450674-92450674 on Build GRCh38
rs758233533 Chr.7: 92450740-92450740 on Build GRCh38

View Full Product Details