Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 48692951-48693216 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IP6K2
Entrez Gene ID
51447
Gene Name
inositol hexakisphosphate kinase 2
Gene Aliases
IHPK2, PIUS
UniGene
Hs.595983
Gene Chromosome Location
Chr.3: 48688003-48717278 on Build GRCh38
Transcripts
NM_001005909.2 NM_016291.3 XM_006713199.3 XM_006713200.1 XM_006713201.1 XM_006713202.1 XM_011533816.1 XM_011533817.1 XM_011533818.1 XM_017006583.1 XM_017006584.1 XM_017006585.1 XM_017006586.1 XM_017006587.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1726314 c.217C>T p.R73C Chr.3: 48693165-48693165 on Build GRCh38
5864756 c.235G>A p.D79N Chr.3: 48693147-48693147 on Build GRCh38
5995097 c.218G>A p.R73H Chr.3: 48693164-48693164 on Build GRCh38
3696134 c.397C>T p.R133C Chr.3: 48692985-48692985 on Build GRCh38
3071135 c.301A>C p.N101H Chr.3: 48693081-48693081 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754220862 Chr.3: 48693085-48693085 on Build GRCh38
rs891026614 Chr.3: 48693143-48693143 on Build GRCh38
rs751006884 Chr.3: 48693047-48693054 on Build GRCh38
rs375195624 Chr.3: 48693165-48693165 on Build GRCh38
rs750673256 Chr.3: 48693003-48693003 on Build GRCh38
rs138179394 Chr.3: 48693157-48693157 on Build GRCh38
rs758817795 Chr.3: 48693136-48693136 on Build GRCh38
rs758750835 Chr.3: 48693048-48693049 on Build GRCh38
rs751060708 Chr.3: 48693065-48693065 on Build GRCh38
rs556681934 Chr.3: 48692977-48692977 on Build GRCh38
rs769023860 Chr.3: 48693164-48693164 on Build GRCh38
rs139352576 Chr.3: 48693020-48693020 on Build GRCh38
rs868659073 Chr.3: 48693119-48693119 on Build GRCh38
rs746328100 Chr.3: 48693135-48693135 on Build GRCh38
rs765975834 Chr.3: 48693055-48693055 on Build GRCh38
rs761045494 Chr.3: 48692992-48692992 on Build GRCh38
rs748666701 Chr.3: 48693000-48693000 on Build GRCh38
rs575264529 Chr.3: 48692985-48692985 on Build GRCh38
rs750310284 Chr.3: 48693034-48693034 on Build GRCh38
rs765704684 Chr.3: 48693001-48693001 on Build GRCh38
rs145148629 Chr.3: 48693037-48693037 on Build GRCh38
rs773276626 Chr.3: 48693152-48693154 on Build GRCh38
rs780321946 Chr.3: 48693154-48693154 on Build GRCh38
rs763325910 Chr.3: 48693083-48693083 on Build GRCh38
rs754662528 Chr.3: 48693066-48693066 on Build GRCh38
rs779291348 Chr.3: 48693118-48693118 on Build GRCh38
rs899611403 Chr.3: 48693177-48693177 on Build GRCh38
rs780448099 Chr.3: 48693055-48693056 on Build GRCh38
rs775587404 Chr.3: 48692984-48692984 on Build GRCh38
rs765120034 Chr.3: 48693012-48693012 on Build GRCh38
rs61754207 Chr.3: 48693117-48693117 on Build GRCh38

View Full Product Details