Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 41440828-41441057 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT38
Entrez Gene ID
8687
Gene Name
keratin 38
Gene Aliases
HA8, KRTHA8, hHa8
UniGene
Hs.248188
Gene Chromosome Location
Chr.17: 41436369-41441344 on Build GRCh38
Transcripts
NM_006771.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4928135 c.1A>G p.M1V Chr.17: 41440921-41440921 on Build GRCh38
6377288 c.14A>G p.Y5C Chr.17: 41440908-41440908 on Build GRCh38
4899731 c.32C>T p.P11L Chr.17: 41440890-41440890 on Build GRCh38
107765 c.79C>T p.P27S Chr.17: 41440843-41440843 on Build GRCh38
2840741 c.18C>T p.S6S Chr.17: 41440904-41440904 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376181996 Chr.17: 41440964-41440964 on Build GRCh38
rs529860068 Chr.17: 41441026-41441026 on Build GRCh38
rs201232784 Chr.17: 41440933-41440933 on Build GRCh38
rs768722264 Chr.17: 41440971-41440971 on Build GRCh38
rs1038583686 Chr.17: 41441034-41441034 on Build GRCh38
rs777509166 Chr.17: 41440943-41440943 on Build GRCh38
rs749337507 Chr.17: 41440898-41440898 on Build GRCh38
rs370945067 Chr.17: 41440851-41440851 on Build GRCh38
rs769897923 Chr.17: 41440885-41440885 on Build GRCh38
rs776449512 Chr.17: 41440973-41440973 on Build GRCh38
rs564129089 Chr.17: 41440927-41440927 on Build GRCh38
rs559394653 Chr.17: 41441014-41441014 on Build GRCh38
rs754474586 Chr.17: 41440920-41440920 on Build GRCh38
rs766036787 Chr.17: 41440913-41440913 on Build GRCh38
rs761233510 Chr.17: 41440911-41440911 on Build GRCh38
rs371151813 Chr.17: 41440946-41440946 on Build GRCh38
rs764804480 Chr.17: 41440922-41440922 on Build GRCh38
rs758125628 Chr.17: 41440860-41440860 on Build GRCh38
rs376974563 Chr.17: 41440906-41440906 on Build GRCh38
rs747928835 Chr.17: 41440853-41440853 on Build GRCh38
rs564553945 Chr.17: 41440961-41440961 on Build GRCh38
rs767975937 Chr.17: 41440878-41440879 on Build GRCh38
rs773923359 Chr.17: 41440908-41440908 on Build GRCh38
rs142601851 Chr.17: 41441027-41441027 on Build GRCh38
rs745621514 Chr.17: 41441021-41441021 on Build GRCh38
rs768790313 Chr.17: 41440902-41440902 on Build GRCh38
rs933927522 Chr.17: 41440917-41440917 on Build GRCh38
rs545926548 Chr.17: 41440907-41440907 on Build GRCh38
rs775405483 Chr.17: 41440895-41440895 on Build GRCh38
rs865853519 Chr.17: 41440901-41440901 on Build GRCh38
rs769748985 Chr.17: 41441025-41441025 on Build GRCh38
rs781140857 Chr.17: 41440959-41440959 on Build GRCh38
rs757205993 Chr.17: 41440948-41440948 on Build GRCh38
rs575311928 Chr.17: 41440903-41440903 on Build GRCh38
rs770653475 Chr.17: 41440876-41440876 on Build GRCh38
rs182943872 Chr.17: 41440982-41440982 on Build GRCh38
rs753311357 Chr.17: 41440918-41440918 on Build GRCh38
rs202240554 Chr.17: 41440864-41440864 on Build GRCh38
rs758250553 Chr.17: 41440935-41440935 on Build GRCh38

View Full Product Details