Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 34370584-34370811 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNE2
Entrez Gene ID
9992
Gene Name
potassium voltage-gated channel subfamily E regulatory subunit 2
Gene Aliases
ATFB4, LQT5, LQT6, MIRP1
UniGene
Hs.551521
Gene Chromosome Location
Chr.21: 34364024-34371141 on Build GRCh38
Transcripts
NM_172201.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3550483 c.280G>A p.E94K Chr.21: 34370758-34370758 on Build GRCh38
6889212 c.331C>A p.H111N Chr.21: 34370809-34370809 on Build GRCh38
3550482 c.240C>A p.S80S Chr.21: 34370718-34370718 on Build GRCh38
3799878 c.268G>C p.E90Q Chr.21: 34370746-34370746 on Build GRCh38
4101427 c.212C>G p.T71S Chr.21: 34370690-34370690 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs961832580 Chr.21: 34370776-34370776 on Build GRCh38
rs74424227 Chr.21: 34370759-34370759 on Build GRCh38
rs771699510 Chr.21: 34370740-34370740 on Build GRCh38
rs1008792812 Chr.21: 34370750-34370750 on Build GRCh38
rs779623946 Chr.21: 34370619-34370619 on Build GRCh38
rs758863768 Chr.21: 34370621-34370621 on Build GRCh38
rs199473649 Chr.21: 34370644-34370644 on Build GRCh38
rs113659353 Chr.21: 34370789-34370789 on Build GRCh38
rs893883306 Chr.21: 34370716-34370716 on Build GRCh38
rs138831412 Chr.21: 34370704-34370704 on Build GRCh38
rs74315448 Chr.21: 34370648-34370648 on Build GRCh38
rs780534713 Chr.21: 34370685-34370685 on Build GRCh38
rs765797117 Chr.21: 34370673-34370673 on Build GRCh38
rs141423405 Chr.21: 34370707-34370707 on Build GRCh38
rs770739581 Chr.21: 34370653-34370653 on Build GRCh38
rs767047359 Chr.21: 34370782-34370782 on Build GRCh38
rs74315447 Chr.21: 34370639-34370639 on Build GRCh38
rs16991654 Chr.21: 34370656-34370656 on Build GRCh38
rs16991656 Chr.21: 34370675-34370675 on Build GRCh38
rs587781005 Chr.21: 34370739-34370739 on Build GRCh38
rs775034771 Chr.21: 34370742-34370742 on Build GRCh38
rs371724328 Chr.21: 34370638-34370638 on Build GRCh38
rs1018893224 Chr.21: 34370752-34370752 on Build GRCh38
rs202169020 Chr.21: 34370706-34370706 on Build GRCh38
rs774193019 Chr.21: 34370724-34370724 on Build GRCh38
rs199473366 Chr.21: 34370747-34370747 on Build GRCh38
rs982620931 Chr.21: 34370632-34370632 on Build GRCh38
rs542835031 Chr.21: 34370683-34370683 on Build GRCh38
rs751276927 Chr.21: 34370621-34370622 on Build GRCh38
rs780568059 Chr.21: 34370774-34370774 on Build GRCh38
rs751891377 Chr.21: 34370786-34370786 on Build GRCh38
rs745605145 Chr.21: 34370674-34370674 on Build GRCh38
rs199473364 Chr.21: 34370671-34370671 on Build GRCh38
rs930268832 Chr.21: 34370664-34370664 on Build GRCh38
rs751931568 Chr.21: 34370687-34370687 on Build GRCh38
rs776764531 Chr.21: 34370634-34370634 on Build GRCh38
rs763545106 Chr.21: 34370781-34370781 on Build GRCh38
rs1028976664 Chr.21: 34370730-34370730 on Build GRCh38
rs767142542 Chr.21: 34370670-34370670 on Build GRCh38
rs763797654 Chr.21: 34370616-34370617 on Build GRCh38
rs199473365 Chr.21: 34370708-34370708 on Build GRCh38
rs889152352 Chr.21: 34370745-34370745 on Build GRCh38
rs748194563 Chr.21: 34370705-34370705 on Build GRCh38
rs1011448773 Chr.21: 34370728-34370728 on Build GRCh38
rs953273203 Chr.21: 34370608-34370608 on Build GRCh38
rs753438833 Chr.21: 34370779-34370779 on Build GRCh38
rs770895197 Chr.21: 34370714-34370714 on Build GRCh38
rs757779830 Chr.21: 34370618-34370618 on Build GRCh38
rs780373385 Chr.21: 34370622-34370622 on Build GRCh38
rs778998757 Chr.21: 34370727-34370727 on Build GRCh38
rs755211128 Chr.21: 34370700-34370700 on Build GRCh38
rs747232176 Chr.21: 34370628-34370628 on Build GRCh38
rs760181970 Chr.21: 34370756-34370756 on Build GRCh38
rs756730160 Chr.21: 34370638-34370639 on Build GRCh38
rs146053586 Chr.21: 34370649-34370649 on Build GRCh38
rs200403369 Chr.21: 34370682-34370682 on Build GRCh38
rs143767851 Chr.21: 34370631-34370631 on Build GRCh38

View Full Product Details