Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 55174677-55174922 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HCRTR2
Entrez Gene ID
3062
Gene Name
hypocretin receptor 2
Gene Aliases
OX2R
UniGene
Hs.151624
Gene Chromosome Location
Chr.6: 55174273-55284698 on Build GRCh38
Transcripts
NM_001526.3 XM_017010798.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1329619 c.99C>G p.D33E Chr.6: 55174686-55174686 on Build GRCh38
176074 c.195C>T p.F65F Chr.6: 55174782-55174782 on Build GRCh38
5561228 c.216C>T p.N72N Chr.6: 55174803-55174803 on Build GRCh38
4689655 c.127C>A p.L43M Chr.6: 55174714-55174714 on Build GRCh38
3628701 c.117C>T p.F39F Chr.6: 55174704-55174704 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759219974 Chr.6: 55174742-55174742 on Build GRCh38
rs779966122 Chr.6: 55174869-55174869 on Build GRCh38
rs752192785 Chr.6: 55174748-55174748 on Build GRCh38
rs780270268 Chr.6: 55174777-55174777 on Build GRCh38
rs781177635 Chr.6: 55174794-55174794 on Build GRCh38
rs995475147 Chr.6: 55174894-55174894 on Build GRCh38
rs539145798 Chr.6: 55174764-55174764 on Build GRCh38
rs764078525 Chr.6: 55174763-55174763 on Build GRCh38
rs939057534 Chr.6: 55174795-55174795 on Build GRCh38
rs200056312 Chr.6: 55174774-55174774 on Build GRCh38
rs930464906 Chr.6: 55174780-55174780 on Build GRCh38
rs368941750 Chr.6: 55174736-55174736 on Build GRCh38
rs773081668 Chr.6: 55174889-55174889 on Build GRCh38
rs373819879 Chr.6: 55174828-55174828 on Build GRCh38
rs1000363903 Chr.6: 55174850-55174850 on Build GRCh38
rs564789310 Chr.6: 55174866-55174866 on Build GRCh38
rs554885516 Chr.6: 55174858-55174858 on Build GRCh38
rs866211132 Chr.6: 55174741-55174741 on Build GRCh38
rs200120685 Chr.6: 55174747-55174747 on Build GRCh38
rs760262376 Chr.6: 55174760-55174760 on Build GRCh38
rs778741456 Chr.6: 55174767-55174767 on Build GRCh38
rs371868747 Chr.6: 55174832-55174832 on Build GRCh38
rs935147099 Chr.6: 55174721-55174721 on Build GRCh38
rs1031806769 Chr.6: 55174864-55174864 on Build GRCh38
rs751758218 Chr.6: 55174860-55174860 on Build GRCh38
rs775309537 Chr.6: 55174824-55174824 on Build GRCh38
rs199972618 Chr.6: 55174857-55174857 on Build GRCh38
rs375022205 Chr.6: 55174826-55174826 on Build GRCh38
rs372213161 Chr.6: 55174696-55174696 on Build GRCh38
rs150493791 Chr.6: 55174800-55174800 on Build GRCh38
rs766548112 Chr.6: 55174859-55174859 on Build GRCh38
rs890663179 Chr.6: 55174839-55174839 on Build GRCh38
rs748656393 Chr.6: 55174796-55174796 on Build GRCh38
rs758195059 Chr.6: 55174853-55174853 on Build GRCh38
rs201306017 Chr.6: 55174709-55174709 on Build GRCh38
rs199728724 Chr.6: 55174695-55174695 on Build GRCh38
rs773609845 Chr.6: 55174820-55174820 on Build GRCh38
rs747025393 Chr.6: 55174786-55174786 on Build GRCh38
rs770172433 Chr.6: 55174716-55174716 on Build GRCh38
rs202246026 Chr.6: 55174745-55174745 on Build GRCh38
rs202040171 Chr.6: 55174852-55174852 on Build GRCh38
rs769661186 Chr.6: 55174888-55174888 on Build GRCh38
rs75924257 Chr.6: 55174789-55174789 on Build GRCh38
rs768754670 Chr.6: 55174791-55174791 on Build GRCh38
rs371585189 Chr.6: 55174838-55174838 on Build GRCh38
rs762698067 Chr.6: 55174898-55174898 on Build GRCh38
rs763420356 Chr.6: 55174822-55174822 on Build GRCh38
rs750255150 Chr.6: 55174769-55174769 on Build GRCh38
rs200553902 Chr.6: 55174701-55174701 on Build GRCh38
rs141403220 Chr.6: 55174766-55174766 on Build GRCh38
rs368082727 Chr.6: 55174849-55174849 on Build GRCh38
rs755232101 Chr.6: 55174862-55174862 on Build GRCh38
rs771779853 Chr.6: 55174823-55174823 on Build GRCh38
rs201269848 Chr.6: 55174835-55174835 on Build GRCh38

View Full Product Details