Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 30890906-30891165 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDR1
Entrez Gene ID
780
Gene Name
discoidin domain receptor tyrosine kinase 1
Gene Aliases
CAK, CD167, DDR, EDDR1, HGK2, MCK10, NEP, NTRK4, PTK3, PTK3A, RTK6, TRKE
UniGene
Hs.631988
Gene Chromosome Location
Chr.6: 30880909-30900156 on Build GRCh38
Transcripts
NM_001202521.1 NM_001202522.1 NM_001202523.1 NM_001297652.1 NM_001297653.1 NM_001297654.1 NM_001954.4 NM_013993.2 NM_013994.2 XM_006715185.2 XM_011514882.2 XM_011514883.1 XM_011514884.1 XM_011514885.1 XM_011514886.1 XM_011514887.1 XM_011514888.2 XM_017011268.1 XM_017011269.1 NR_039783.1

Gene Symbol
MIR4640
Entrez Gene ID
100616237
Gene Name
microRNA 4640
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.6: 30890883-30890972 on Build GRCh38
Transcripts
NM_001202521.1 NM_001202522.1 NM_001202523.1 NM_001297652.1 NM_001297653.1 NM_001297654.1 NM_001954.4 NM_013993.2 NM_013994.2 XM_006715185.2 XM_011514882.2 XM_011514883.1 XM_011514884.1 XM_011514885.1 XM_011514886.1 XM_011514887.1 XM_011514888.2 XM_017011268.1 XM_017011269.1 NR_039783.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1173212 c.511G>C p.D171H Chr.6: 30891066-30891066 on Build GRCh38
5524820 c.444G>A p.E148E Chr.6: 30890999-30890999 on Build GRCh38
1329097 c.466delG p.M159fs*16 Chr.6: 30891021-30891021 on Build GRCh38
3861548 c.443A>G p.E148G Chr.6: 30890998-30890998 on Build GRCh38
7142254 c.522G>T p.M174I Chr.6: 30891077-30891077 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766274112 Chr.6: 30891023-30891024 on Build GRCh38
rs749224278 Chr.6: 30891028-30891028 on Build GRCh38
rs777826215 Chr.6: 30890997-30890997 on Build GRCh38
rs1011484740 Chr.6: 30891057-30891057 on Build GRCh38
rs553030737 Chr.6: 30890936-30890936 on Build GRCh38
rs138528891 Chr.6: 30890937-30890937 on Build GRCh38
rs139811882 Chr.6: 30890992-30890992 on Build GRCh38
rs779781546 Chr.6: 30891023-30891023 on Build GRCh38
rs749065685 Chr.6: 30891116-30891116 on Build GRCh38
rs936816885 Chr.6: 30891018-30891018 on Build GRCh38
rs761229007 Chr.6: 30891040-30891040 on Build GRCh38
rs1022417101 Chr.6: 30891060-30891060 on Build GRCh38
rs963227228 Chr.6: 30891131-30891131 on Build GRCh38
rs753667209 Chr.6: 30890941-30890941 on Build GRCh38
rs55980643 Chr.6: 30891061-30891061 on Build GRCh38
rs544616376 Chr.6: 30890948-30890948 on Build GRCh38
rs367606595 Chr.6: 30891056-30891056 on Build GRCh38
rs1049622 Chr.6: 30891080-30891080 on Build GRCh38
rs746554342 Chr.6: 30891111-30891111 on Build GRCh38
rs144609719 Chr.6: 30891021-30891021 on Build GRCh38
rs141547665 Chr.6: 30891069-30891069 on Build GRCh38
rs745723181 Chr.6: 30891010-30891010 on Build GRCh38
rs149184910 Chr.6: 30891121-30891121 on Build GRCh38
rs757401499 Chr.6: 30891110-30891110 on Build GRCh38
rs61596908 Chr.6: 30890955-30890956 on Build GRCh38
rs755503620 Chr.6: 30890961-30890961 on Build GRCh38
rs750870524 Chr.6: 30890939-30890939 on Build GRCh38
rs765835603 Chr.6: 30891079-30891079 on Build GRCh38
rs768519318 Chr.6: 30891036-30891036 on Build GRCh38
rs374498140 Chr.6: 30891144-30891144 on Build GRCh38
rs267600944 Chr.6: 30891038-30891038 on Build GRCh38
rs745485473 Chr.6: 30891049-30891049 on Build GRCh38
rs949637145 Chr.6: 30890951-30890951 on Build GRCh38
rs773912777 Chr.6: 30891037-30891037 on Build GRCh38
rs549249224 Chr.6: 30891083-30891083 on Build GRCh38
rs779138842 Chr.6: 30890962-30890962 on Build GRCh38
rs564497322 Chr.6: 30891044-30891044 on Build GRCh38
rs918173642 Chr.6: 30890950-30890950 on Build GRCh38
rs544509367 Chr.6: 30890956-30890956 on Build GRCh38
rs754189500 Chr.6: 30891070-30891070 on Build GRCh38
rs751759098 Chr.6: 30891090-30891090 on Build GRCh38
rs1004790110 Chr.6: 30891103-30891103 on Build GRCh38
rs56231803 Chr.6: 30891064-30891064 on Build GRCh38
rs145618831 Chr.6: 30891071-30891071 on Build GRCh38
rs758384854 Chr.6: 30891081-30891081 on Build GRCh38
rs758712984 Chr.6: 30890993-30890993 on Build GRCh38
rs778795515 Chr.6: 30891126-30891126 on Build GRCh38

View Full Product Details