Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 70657132-70657395 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JCHAIN
Entrez Gene ID
3512
Gene Name
joining chain of multimeric IgA and IgM
Gene Aliases
IGCJ, IGJ, JCH
UniGene
Hs.643431
Gene Chromosome Location
Chr.4: 70655541-70676901 on Build GRCh38
Transcripts
NM_144646.3 XM_006714211.2 XM_006714212.2 XM_011531925.1 XM_011531926.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4988610 c.233C>T p.P78L Chr.4: 70657247-70657247 on Build GRCh38
3775957 c.216C>A p.I72I Chr.4: 70657264-70657264 on Build GRCh38
6197192 c.263C>T p.S88F Chr.4: 70657217-70657217 on Build GRCh38
1430810 c.239G>T p.R80I Chr.4: 70657241-70657241 on Build GRCh38
5017090 c.213T>A p.N71K Chr.4: 70657267-70657267 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs576956923 Chr.4: 70657191-70657191 on Build GRCh38
rs370304781 Chr.4: 70657174-70657174 on Build GRCh38
rs760158502 Chr.4: 70657194-70657194 on Build GRCh38
rs753528709 Chr.4: 70657184-70657184 on Build GRCh38
rs533905418 Chr.4: 70657177-70657177 on Build GRCh38
rs114837274 Chr.4: 70657351-70657351 on Build GRCh38
rs774719114 Chr.4: 70657248-70657248 on Build GRCh38
rs746006842 Chr.4: 70657202-70657202 on Build GRCh38
rs772161700 Chr.4: 70657198-70657198 on Build GRCh38
rs910591847 Chr.4: 70657344-70657344 on Build GRCh38
rs749978722 Chr.4: 70657175-70657175 on Build GRCh38
rs924730867 Chr.4: 70657268-70657268 on Build GRCh38
rs779806926 Chr.4: 70657179-70657179 on Build GRCh38
rs75928327 Chr.4: 70657282-70657282 on Build GRCh38
rs34204602 Chr.4: 70657173-70657174 on Build GRCh38
rs367674123 Chr.4: 70657336-70657336 on Build GRCh38
rs886638618 Chr.4: 70657212-70657212 on Build GRCh38
rs542037538 Chr.4: 70657360-70657360 on Build GRCh38
rs181994126 Chr.4: 70657291-70657291 on Build GRCh38
rs762123949 Chr.4: 70657261-70657261 on Build GRCh38
rs34761034 Chr.4: 70657363-70657364 on Build GRCh38
rs775844392 Chr.4: 70657235-70657235 on Build GRCh38
rs1051921185 Chr.4: 70657190-70657190 on Build GRCh38
rs201053863 Chr.4: 70657241-70657241 on Build GRCh38
rs768807937 Chr.4: 70657247-70657247 on Build GRCh38
rs1037896113 Chr.4: 70657286-70657286 on Build GRCh38
rs112978189 Chr.4: 70657292-70657292 on Build GRCh38
rs6831615 Chr.4: 70657364-70657364 on Build GRCh38
rs1045510114 Chr.4: 70657363-70657363 on Build GRCh38
rs754694526 Chr.4: 70657341-70657341 on Build GRCh38
rs1022165401 Chr.4: 70657170-70657170 on Build GRCh38
rs765802261 Chr.4: 70657316-70657316 on Build GRCh38
rs763162273 Chr.4: 70657238-70657238 on Build GRCh38
rs769745995 Chr.4: 70657222-70657222 on Build GRCh38
rs558574156 Chr.4: 70657188-70657188 on Build GRCh38
rs745896881 Chr.4: 70657245-70657245 on Build GRCh38
rs768111301 Chr.4: 70657272-70657272 on Build GRCh38
rs763651633 Chr.4: 70657193-70657194 on Build GRCh38

View Full Product Details