Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 18229898-18230147 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LLGL1
Entrez Gene ID
3996
Gene Name
LLGL1, scribble cell polarity complex component
Gene Aliases
DLG4, HUGL, HUGL-1, HUGL1, LLGL, Lgl1, Mgl1
UniGene
Hs.513983
Gene Chromosome Location
Chr.17: 18225593-18244875 on Build GRCh38
Transcripts
NM_004140.3 XM_011523849.2 XM_011523850.2 XM_011523851.2 XM_011523852.2 XM_011523853.2 XM_011523854.2 XM_011523855.2 XM_011523856.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7148367 c.153C>T p.I51I Chr.17: 18230012-18230012 on Build GRCh38
7190077 c.92A>G p.H31R Chr.17: 18229951-18229951 on Build GRCh38
6080424 c.167G>T p.G56V Chr.17: 18230026-18230026 on Build GRCh38
7315635 c.88G>A p.E30K Chr.17: 18229947-18229947 on Build GRCh38
4465777 c.139C>T p.R47C Chr.17: 18229998-18229998 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760918366 Chr.17: 18229980-18229980 on Build GRCh38
rs779589211 Chr.17: 18230027-18230027 on Build GRCh38
rs540140275 Chr.17: 18229998-18229998 on Build GRCh38
rs761921235 Chr.17: 18229991-18229991 on Build GRCh38
rs766097233 Chr.17: 18230005-18230005 on Build GRCh38
rs755632435 Chr.17: 18230021-18230021 on Build GRCh38
rs772573509 Chr.17: 18230064-18230064 on Build GRCh38
rs1008712739 Chr.17: 18230124-18230124 on Build GRCh38
rs996078565 Chr.17: 18230094-18230094 on Build GRCh38
rs759475436 Chr.17: 18229936-18229937 on Build GRCh38
rs774177410 Chr.17: 18229927-18229928 on Build GRCh38
rs368206803 Chr.17: 18229999-18229999 on Build GRCh38
rs73288285 Chr.17: 18229930-18229930 on Build GRCh38
rs769876442 Chr.17: 18230080-18230080 on Build GRCh38
rs772520170 Chr.17: 18229973-18229973 on Build GRCh38
rs553674726 Chr.17: 18230065-18230065 on Build GRCh38
rs183699036 Chr.17: 18230066-18230066 on Build GRCh38
rs372502097 Chr.17: 18230090-18230090 on Build GRCh38
rs776887896 Chr.17: 18230073-18230073 on Build GRCh38
rs890392904 Chr.17: 18230095-18230095 on Build GRCh38
rs755402220 Chr.17: 18229923-18229923 on Build GRCh38
rs977234252 Chr.17: 18229970-18229970 on Build GRCh38
rs780512571 Chr.17: 18229921-18229921 on Build GRCh38
rs145857913 Chr.17: 18230012-18230012 on Build GRCh38
rs141705015 Chr.17: 18229990-18229990 on Build GRCh38
rs115967277 Chr.17: 18229963-18229963 on Build GRCh38
rs746307176 Chr.17: 18230035-18230035 on Build GRCh38
rs768908181 Chr.17: 18229985-18229985 on Build GRCh38
rs748199403 Chr.17: 18230076-18230076 on Build GRCh38
rs773139416 Chr.17: 18230081-18230081 on Build GRCh38
rs899876098 Chr.17: 18230084-18230084 on Build GRCh38
rs752191168 Chr.17: 18230013-18230013 on Build GRCh38
rs372102209 Chr.17: 18229938-18229938 on Build GRCh38
rs766939885 Chr.17: 18229943-18229943 on Build GRCh38
rs751226368 Chr.17: 18230010-18230010 on Build GRCh38
rs376398557 Chr.17: 18229974-18229974 on Build GRCh38
rs1015175130 Chr.17: 18230127-18230127 on Build GRCh38
rs369183356 Chr.17: 18229982-18229982 on Build GRCh38

View Full Product Details