Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67940166-67940407 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LCAT
Entrez Gene ID
3931
Gene Name
lecithin-cholesterol acyltransferase
Gene Aliases
-
UniGene
Hs.387239
Gene Chromosome Location
Chr.16: 67939884-67944112 on Build GRCh38
Transcripts
NM_000229.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
972630 c.840C>T p.R280R Chr.16: 67940387-67940387 on Build GRCh38
972631 c.839G>A p.R280H Chr.16: 67940388-67940388 on Build GRCh38
1609539 c.1048A>C p.I350L Chr.16: 67940179-67940179 on Build GRCh38
6195206 c.824G>C p.W275S Chr.16: 67940403-67940403 on Build GRCh38
972628 c.973C>T p.L325L Chr.16: 67940254-67940254 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373437836 Chr.16: 67940387-67940387 on Build GRCh38
rs948776335 Chr.16: 67940243-67940243 on Build GRCh38
rs767508071 Chr.16: 67940259-67940259 on Build GRCh38
rs767628388 Chr.16: 67940366-67940366 on Build GRCh38
rs765119491 Chr.16: 67940381-67940381 on Build GRCh38
rs368229427 Chr.16: 67940187-67940187 on Build GRCh38
rs758221629 Chr.16: 67940291-67940291 on Build GRCh38
rs200225997 Chr.16: 67940348-67940348 on Build GRCh38
rs746487870 Chr.16: 67940206-67940206 on Build GRCh38
rs138943456 Chr.16: 67940210-67940210 on Build GRCh38
rs757228770 Chr.16: 67940371-67940371 on Build GRCh38
rs752443204 Chr.16: 67940290-67940290 on Build GRCh38
rs952228539 Chr.16: 67940255-67940255 on Build GRCh38
rs753759978 Chr.16: 67940262-67940262 on Build GRCh38
rs1046996034 Chr.16: 67940326-67940326 on Build GRCh38
rs121908048 Chr.16: 67940276-67940276 on Build GRCh38
rs112299487 Chr.16: 67940209-67940209 on Build GRCh38
rs750551944 Chr.16: 67940367-67940367 on Build GRCh38
rs761999436 Chr.16: 67940360-67940360 on Build GRCh38
rs28940888 Chr.16: 67940193-67940193 on Build GRCh38
rs202017590 Chr.16: 67940188-67940188 on Build GRCh38
rs770469492 Chr.16: 67940328-67940328 on Build GRCh38
rs770281333 Chr.16: 67940222-67940222 on Build GRCh38
rs780635568 Chr.16: 67940324-67940324 on Build GRCh38
rs139453193 Chr.16: 67940246-67940246 on Build GRCh38
rs770356071 Chr.16: 67940207-67940207 on Build GRCh38
rs753873629 Chr.16: 67940198-67940199 on Build GRCh38
rs763496682 Chr.16: 67940187-67940188 on Build GRCh38
rs776035233 Chr.16: 67940230-67940230 on Build GRCh38
rs747769382 Chr.16: 67940194-67940194 on Build GRCh38
rs149389101 Chr.16: 67940302-67940302 on Build GRCh38
rs995369011 Chr.16: 67940300-67940300 on Build GRCh38
rs763241601 Chr.16: 67940239-67940239 on Build GRCh38
rs959668537 Chr.16: 67940357-67940357 on Build GRCh38
rs777417745 Chr.16: 67940293-67940293 on Build GRCh38
rs996523378 Chr.16: 67940354-67940354 on Build GRCh38
rs778658721 Chr.16: 67940278-67940278 on Build GRCh38
rs5924 Chr.16: 67940327-67940327 on Build GRCh38
rs761884788 Chr.16: 67940251-67940251 on Build GRCh38
rs772762775 Chr.16: 67940200-67940200 on Build GRCh38
rs763366383 Chr.16: 67940334-67940334 on Build GRCh38
rs559100013 Chr.16: 67940329-67940329 on Build GRCh38
rs532898663 Chr.16: 67940388-67940388 on Build GRCh38
rs121908056 Chr.16: 67940255-67940257 on Build GRCh38
rs754974345 Chr.16: 67940277-67940277 on Build GRCh38
rs749186904 Chr.16: 67940192-67940192 on Build GRCh38
rs771761885 Chr.16: 67940196-67940196 on Build GRCh38
rs760768263 Chr.16: 67940375-67940375 on Build GRCh38
rs375740669 Chr.16: 67940212-67940212 on Build GRCh38
rs765744503 Chr.16: 67940233-67940233 on Build GRCh38
rs752637398 Chr.16: 67940386-67940386 on Build GRCh38
rs1019658248 Chr.16: 67940336-67940336 on Build GRCh38
rs774574304 Chr.16: 67940359-67940359 on Build GRCh38
rs751757278 Chr.16: 67940247-67940247 on Build GRCh38

View Full Product Details