Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 73434382-73434631 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FZD9
Entrez Gene ID
8326
Gene Name
frizzled class receptor 9
Gene Aliases
CD349, FZD3
UniGene
Hs.647029
Gene Chromosome Location
Chr.7: 73433779-73436120 on Build GRCh38
Transcripts
NM_003508.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4421612 c.393C>T p.F131F Chr.7: 73434400-73434400 on Build GRCh38
4685997 c.450G>A p.A150A Chr.7: 73434457-73434457 on Build GRCh38
1091547 c.589G>A p.E197K Chr.7: 73434596-73434596 on Build GRCh38
4685996 c.432G>A p.T144T Chr.7: 73434439-73434439 on Build GRCh38
6226431 c.431C>T p.T144M Chr.7: 73434438-73434438 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769369970 Chr.7: 73434573-73434573 on Build GRCh38
rs368682922 Chr.7: 73434409-73434409 on Build GRCh38
rs781938252 Chr.7: 73434592-73434592 on Build GRCh38
rs781853577 Chr.7: 73434605-73434605 on Build GRCh38
rs782702023 Chr.7: 73434437-73434437 on Build GRCh38
rs536929629 Chr.7: 73434524-73434524 on Build GRCh38
rs575621716 Chr.7: 73434582-73434582 on Build GRCh38
rs782330628 Chr.7: 73434506-73434506 on Build GRCh38
rs782073205 Chr.7: 73434405-73434405 on Build GRCh38
rs201167285 Chr.7: 73434475-73434475 on Build GRCh38
rs782795346 Chr.7: 73434610-73434610 on Build GRCh38
rs782136427 Chr.7: 73434606-73434606 on Build GRCh38
rs781972322 Chr.7: 73434408-73434408 on Build GRCh38
rs782300865 Chr.7: 73434581-73434581 on Build GRCh38
rs538778244 Chr.7: 73434419-73434419 on Build GRCh38
rs782077630 Chr.7: 73434602-73434602 on Build GRCh38
rs17852398 Chr.7: 73434603-73434603 on Build GRCh38
rs782351404 Chr.7: 73434406-73434406 on Build GRCh38
rs138414411 Chr.7: 73434563-73434563 on Build GRCh38
rs782760906 Chr.7: 73434604-73434604 on Build GRCh38
rs782384125 Chr.7: 73434547-73434547 on Build GRCh38
rs782180751 Chr.7: 73434505-73434505 on Build GRCh38
rs782636147 Chr.7: 73434441-73434441 on Build GRCh38
rs782047303 Chr.7: 73434428-73434428 on Build GRCh38
rs782418729 Chr.7: 73434402-73434402 on Build GRCh38
rs781871788 Chr.7: 73434454-73434454 on Build GRCh38
rs570248551 Chr.7: 73434529-73434529 on Build GRCh38
rs372342933 Chr.7: 73434424-73434424 on Build GRCh38
rs202012985 Chr.7: 73434439-73434439 on Build GRCh38
rs782541209 Chr.7: 73434457-73434457 on Build GRCh38
rs782692525 Chr.7: 73434497-73434497 on Build GRCh38

View Full Product Details