Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 43643329-43643566 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MEOX1
Entrez Gene ID
4222
Gene Name
mesenchyme homeobox 1
Gene Aliases
KFS2, MOX1
UniGene
Hs.438
Gene Chromosome Location
Chr.17: 43640389-43661954 on Build GRCh38
Transcripts
NM_001040002.1 NM_004527.3 NM_013999.3 XM_011524818.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5138350 c.637C>T p.R213C Chr.17: 43643493-43643493 on Build GRCh38
4066805 c.620A>G p.N207S Chr.17: 43643510-43643510 on Build GRCh38
6711453 c.615G>A p.A205A Chr.17: 43643515-43643515 on Build GRCh38
4066806 c.592C>T p.R198W Chr.17: 43643538-43643538 on Build GRCh38
979784 c.599G>A p.R200H Chr.17: 43643531-43643531 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs981653946 Chr.17: 43643533-43643533 on Build GRCh38
rs778513929 Chr.17: 43643515-43643515 on Build GRCh38
rs780707649 Chr.17: 43643532-43643532 on Build GRCh38
rs371698055 Chr.17: 43643465-43643465 on Build GRCh38
rs769494867 Chr.17: 43643538-43643538 on Build GRCh38
rs1015094658 Chr.17: 43643497-43643497 on Build GRCh38
rs755249945 Chr.17: 43643507-43643507 on Build GRCh38
rs963703606 Chr.17: 43643467-43643468 on Build GRCh38
rs986083481 Chr.17: 43643389-43643389 on Build GRCh38
rs772847956 Chr.17: 43643458-43643460 on Build GRCh38
rs776306534 Chr.17: 43643466-43643466 on Build GRCh38
rs975200690 Chr.17: 43643438-43643438 on Build GRCh38
rs528953630 Chr.17: 43643430-43643430 on Build GRCh38
rs375183146 Chr.17: 43643477-43643477 on Build GRCh38
rs777876074 Chr.17: 43643477-43643478 on Build GRCh38
rs893251387 Chr.17: 43643542-43643542 on Build GRCh38
rs138317571 Chr.17: 43643484-43643484 on Build GRCh38
rs746582127 Chr.17: 43643463-43643463 on Build GRCh38
rs747977868 Chr.17: 43643516-43643516 on Build GRCh38
rs931057527 Chr.17: 43643460-43643460 on Build GRCh38
rs903075882 Chr.17: 43643435-43643435 on Build GRCh38
rs772059036 Chr.17: 43643444-43643444 on Build GRCh38
rs967639266 Chr.17: 43643357-43643357 on Build GRCh38
rs758257527 Chr.17: 43643518-43643518 on Build GRCh38
rs1055805985 Chr.17: 43643374-43643374 on Build GRCh38
rs747985182 Chr.17: 43643441-43643441 on Build GRCh38
rs746492281 Chr.17: 43643524-43643524 on Build GRCh38
rs986981225 Chr.17: 43643375-43643375 on Build GRCh38
rs766674093 Chr.17: 43643496-43643496 on Build GRCh38
rs368246577 Chr.17: 43643481-43643481 on Build GRCh38
rs983058690 Chr.17: 43643468-43643468 on Build GRCh38
rs772527885 Chr.17: 43643453-43643453 on Build GRCh38
rs754113524 Chr.17: 43643509-43643509 on Build GRCh38
rs764320690 Chr.17: 43643478-43643478 on Build GRCh38
rs544228550 Chr.17: 43643475-43643475 on Build GRCh38
rs1020449881 Chr.17: 43643355-43643355 on Build GRCh38
rs914444862 Chr.17: 43643420-43643420 on Build GRCh38
rs762635938 Chr.17: 43643478-43643478 on Build GRCh38
rs189575498 Chr.17: 43643416-43643416 on Build GRCh38
rs199877643 Chr.17: 43643485-43643485 on Build GRCh38
rs745469637 Chr.17: 43643537-43643537 on Build GRCh38
rs754073060 Chr.17: 43643498-43643498 on Build GRCh38
rs1011811774 Chr.17: 43643400-43643400 on Build GRCh38
rs375775745 Chr.17: 43643483-43643483 on Build GRCh38
rs1019244745 Chr.17: 43643414-43643414 on Build GRCh38

View Full Product Details