Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 206969039-206969286 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPO
Entrez Gene ID
130749
Gene Name
carboxypeptidase O
Gene Aliases
-
UniGene
Hs.684103
Gene Chromosome Location
Chr.2: 206932853-206971064 on Build GRCh38
Transcripts
NM_173077.2 XM_017003372.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1631771 c.946A>T p.T316S Chr.2: 206969257-206969257 on Build GRCh38
3991004 c.933G>C p.L311L Chr.2: 206969244-206969244 on Build GRCh38
4583116 c.866C>T p.A289V Chr.2: 206969177-206969177 on Build GRCh38
7362504 c.898G>A p.D300N Chr.2: 206969209-206969209 on Build GRCh38
387207 c.912C>A p.P304P Chr.2: 206969223-206969223 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745904494 Chr.2: 206969235-206969235 on Build GRCh38
rs917282531 Chr.2: 206969222-206969222 on Build GRCh38
rs890356469 Chr.2: 206969109-206969109 on Build GRCh38
rs747443946 Chr.2: 206969207-206969207 on Build GRCh38
rs371700076 Chr.2: 206969192-206969192 on Build GRCh38
rs769941479 Chr.2: 206969248-206969248 on Build GRCh38
rs7582092 Chr.2: 206969124-206969124 on Build GRCh38
rs192508378 Chr.2: 206969245-206969245 on Build GRCh38
rs1008800073 Chr.2: 206969116-206969116 on Build GRCh38
rs7582305 Chr.2: 206969259-206969259 on Build GRCh38
rs754559988 Chr.2: 206969180-206969180 on Build GRCh38
rs368130794 Chr.2: 206969170-206969170 on Build GRCh38
rs767358577 Chr.2: 206969148-206969148 on Build GRCh38
rs1047887674 Chr.2: 206969068-206969068 on Build GRCh38
rs753002825 Chr.2: 206969131-206969131 on Build GRCh38
rs971610651 Chr.2: 206969247-206969247 on Build GRCh38
rs758629422 Chr.2: 206969203-206969203 on Build GRCh38
rs980367519 Chr.2: 206969250-206969250 on Build GRCh38
rs773795517 Chr.2: 206969184-206969193 on Build GRCh38
rs547219145 Chr.2: 206969147-206969147 on Build GRCh38
rs1023013200 Chr.2: 206969067-206969067 on Build GRCh38
rs772982004 Chr.2: 206969264-206969264 on Build GRCh38
rs964534233 Chr.2: 206969138-206969138 on Build GRCh38
rs756045381 Chr.2: 206969161-206969161 on Build GRCh38
rs993169072 Chr.2: 206969065-206969065 on Build GRCh38
rs202095741 Chr.2: 206969253-206969253 on Build GRCh38
rs747856779 Chr.2: 206969199-206969199 on Build GRCh38
rs149624912 Chr.2: 206969187-206969187 on Build GRCh38
rs771550956 Chr.2: 206969216-206969216 on Build GRCh38
rs1033107037 Chr.2: 206969176-206969176 on Build GRCh38
rs374554316 Chr.2: 206969234-206969234 on Build GRCh38
rs570745087 Chr.2: 206969159-206969159 on Build GRCh38
rs770199489 Chr.2: 206969115-206969115 on Build GRCh38
rs763241786 Chr.2: 206969258-206969258 on Build GRCh38
rs759125914 Chr.2: 206969177-206969177 on Build GRCh38
rs112868011 Chr.2: 206969206-206969206 on Build GRCh38
rs776846026 Chr.2: 206969123-206969123 on Build GRCh38

View Full Product Details