Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 31290768-31291003 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIK3IP1
Entrez Gene ID
113791
Gene Name
phosphoinositide-3-kinase interacting protein 1
Gene Aliases
HGFL, hHGFL(S)
UniGene
Hs.26670
Gene Chromosome Location
Chr.22: 31281593-31292534 on Build GRCh38
Transcripts
NM_001135911.1 NM_052880.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5859200 c.307G>A p.E103K Chr.22: 31290965-31290965 on Build GRCh38
5859199 c.307G>A p.E103K Chr.22: 31290965-31290965 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs938878318 Chr.22: 31290866-31290866 on Build GRCh38
rs191721373 Chr.22: 31290937-31290937 on Build GRCh38
rs757129559 Chr.22: 31290932-31290932 on Build GRCh38
rs750268792 Chr.22: 31290984-31290984 on Build GRCh38
rs561070833 Chr.22: 31290913-31290913 on Build GRCh38
rs898183652 Chr.22: 31290882-31290882 on Build GRCh38
rs1025322680 Chr.22: 31290806-31290806 on Build GRCh38
rs942633627 Chr.22: 31290824-31290824 on Build GRCh38
rs1008519388 Chr.22: 31290911-31290911 on Build GRCh38
rs761940617 Chr.22: 31290960-31290960 on Build GRCh38
rs972468516 Chr.22: 31290788-31290790 on Build GRCh38
rs1019486069 Chr.22: 31290862-31290862 on Build GRCh38
rs990080251 Chr.22: 31290945-31290945 on Build GRCh38
rs775514709 Chr.22: 31290953-31290953 on Build GRCh38
rs774698844 Chr.22: 31290957-31290957 on Build GRCh38
rs956897201 Chr.22: 31290976-31290976 on Build GRCh38
rs1042964678 Chr.22: 31290828-31290828 on Build GRCh38
rs563707948 Chr.22: 31290947-31290947 on Build GRCh38
rs901252081 Chr.22: 31290879-31290879 on Build GRCh38
rs928174562 Chr.22: 31290964-31290964 on Build GRCh38
rs947079743 Chr.22: 31290827-31290827 on Build GRCh38
rs763139396 Chr.22: 31290954-31290954 on Build GRCh38
rs370397734 Chr.22: 31290928-31290928 on Build GRCh38
rs988932134 Chr.22: 31290977-31290977 on Build GRCh38
rs775567949 Chr.22: 31290914-31290914 on Build GRCh38
rs574670234 Chr.22: 31290919-31290919 on Build GRCh38
rs747322782 Chr.22: 31290939-31290939 on Build GRCh38
rs1006707384 Chr.22: 31290861-31290861 on Build GRCh38
rs767366392 Chr.22: 31290983-31290983 on Build GRCh38
rs894596100 Chr.22: 31290872-31290872 on Build GRCh38
rs1025841970 Chr.22: 31290891-31290891 on Build GRCh38
rs750315483 Chr.22: 31290934-31290934 on Build GRCh38
rs914468154 Chr.22: 31290958-31290958 on Build GRCh38
rs572301134 Chr.22: 31290886-31290886 on Build GRCh38
rs1025731833 Chr.22: 31290887-31290887 on Build GRCh38
rs771352902 Chr.22: 31290944-31290944 on Build GRCh38
rs770316470 Chr.22: 31290950-31290950 on Build GRCh38
rs750010168 Chr.22: 31290924-31290925 on Build GRCh38
rs1006435109 Chr.22: 31290874-31290874 on Build GRCh38
rs765969686 Chr.22: 31290969-31290970 on Build GRCh38
rs755758978 Chr.22: 31290954-31290955 on Build GRCh38
rs946003376 Chr.22: 31290834-31290834 on Build GRCh38
rs536394940 Chr.22: 31290854-31290854 on Build GRCh38
rs912992540 Chr.22: 31290980-31290980 on Build GRCh38
rs767554256 Chr.22: 31290933-31290933 on Build GRCh38
rs141171195 Chr.22: 31290823-31290823 on Build GRCh38
rs541213034 Chr.22: 31290894-31290894 on Build GRCh38
rs955404990 Chr.22: 31290814-31290814 on Build GRCh38
rs746324699 Chr.22: 31290948-31290948 on Build GRCh38
rs986636369 Chr.22: 31290821-31290821 on Build GRCh38
rs1055964546 Chr.22: 31290867-31290867 on Build GRCh38
rs764206212 Chr.22: 31290955-31290955 on Build GRCh38
rs779665421 Chr.22: 31290936-31290936 on Build GRCh38
rs1052784484 Chr.22: 31290807-31290807 on Build GRCh38
rs141504589 Chr.22: 31290985-31290985 on Build GRCh38

View Full Product Details