Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7497935-7498166 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POLR2A
Entrez Gene ID
5430
Gene Name
RNA polymerase II subunit A
Gene Aliases
POLR2, POLRA, RPB1, RPBh1, RPO2, RPOL2, RpIILS, hRPB220, hsRPB1
UniGene
Hs.270017
Gene Chromosome Location
Chr.17: 7484379-7514618 on Build GRCh38
Transcripts
NM_000937.4

SNP Detail

SNP ID Chromosome Location
rs886534799 Chr.17: 7498133-7498133 on Build GRCh38
rs763272826 Chr.17: 7498121-7498121 on Build GRCh38
rs1049032120 Chr.17: 7498126-7498126 on Build GRCh38
rs988954985 Chr.17: 7498037-7498037 on Build GRCh38
rs1038961104 Chr.17: 7498062-7498062 on Build GRCh38
rs925250302 Chr.17: 7498107-7498107 on Build GRCh38
rs761744418 Chr.17: 7498114-7498114 on Build GRCh38
rs755527636 Chr.17: 7498052-7498052 on Build GRCh38
rs759589337 Chr.17: 7498034-7498034 on Build GRCh38
rs372468225 Chr.17: 7498105-7498105 on Build GRCh38
rs961145159 Chr.17: 7497960-7497960 on Build GRCh38
rs752028574 Chr.17: 7498051-7498051 on Build GRCh38
rs765310036 Chr.17: 7498041-7498041 on Build GRCh38
rs141769858 Chr.17: 7498095-7498095 on Build GRCh38
rs916633343 Chr.17: 7498123-7498123 on Build GRCh38
rs776911914 Chr.17: 7498038-7498038 on Build GRCh38
rs758643684 Chr.17: 7498047-7498047 on Build GRCh38
rs987372651 Chr.17: 7498094-7498094 on Build GRCh38
rs966918595 Chr.17: 7498098-7498098 on Build GRCh38
rs368653926 Chr.17: 7498035-7498035 on Build GRCh38
rs969721482 Chr.17: 7498010-7498010 on Build GRCh38
rs777229954 Chr.17: 7498073-7498073 on Build GRCh38
rs780666921 Chr.17: 7498056-7498056 on Build GRCh38
rs751974951 Chr.17: 7498136-7498136 on Build GRCh38
rs371165806 Chr.17: 7498031-7498031 on Build GRCh38
rs777112017 Chr.17: 7498034-7498035 on Build GRCh38
rs936751827 Chr.17: 7498111-7498111 on Build GRCh38
rs769391040 Chr.17: 7498027-7498027 on Build GRCh38
rs752211472 Chr.17: 7498058-7498058 on Build GRCh38
rs750672343 Chr.17: 7498044-7498044 on Build GRCh38
rs201604230 Chr.17: 7498025-7498025 on Build GRCh38
rs778903563 Chr.17: 7498096-7498096 on Build GRCh38
rs1054982471 Chr.17: 7497963-7497963 on Build GRCh38
rs748989665 Chr.17: 7498078-7498078 on Build GRCh38
rs886350259 Chr.17: 7498021-7498021 on Build GRCh38
rs199991956 Chr.17: 7498016-7498016 on Build GRCh38
rs368088717 Chr.17: 7498029-7498029 on Build GRCh38
rs745789004 Chr.17: 7498102-7498102 on Build GRCh38
rs181418373 Chr.17: 7498018-7498018 on Build GRCh38
rs1023887400 Chr.17: 7497969-7497969 on Build GRCh38
rs769941878 Chr.17: 7498120-7498120 on Build GRCh38
rs748699394 Chr.17: 7498014-7498014 on Build GRCh38
rs755650853 Chr.17: 7498070-7498070 on Build GRCh38
rs75930136 Chr.17: 7498040-7498040 on Build GRCh38
rs766762863 Chr.17: 7498048-7498048 on Build GRCh38
rs576600063 Chr.17: 7498011-7498011 on Build GRCh38
rs373855132 Chr.17: 7498032-7498032 on Build GRCh38
rs554698045 Chr.17: 7498009-7498009 on Build GRCh38
rs1012544865 Chr.17: 7497962-7497962 on Build GRCh38
rs760028176 Chr.17: 7498141-7498141 on Build GRCh38
rs25603 Chr.17: 7498017-7498018 on Build GRCh38
rs766705311 Chr.17: 7498125-7498125 on Build GRCh38
rs772202966 Chr.17: 7498104-7498104 on Build GRCh38
rs111845660 Chr.17: 7498016-7498017 on Build GRCh38

View Full Product Details