Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 46260392-46260647 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAD54L
Entrez Gene ID
8438
Gene Name
RAD54-like (S. cerevisiae)
Gene Aliases
HR54, RAD54A, hHR54, hRAD54
UniGene
Hs.642042
Gene Chromosome Location
Chr.1: 46247695-46278477 on Build GRCh38
Transcripts
NM_001142548.1 NM_003579.3 XM_006710975.3 XM_011542299.2 XM_011542300.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6288168 c.421G>T p.V141F Chr.1: 46260555-46260555 on Build GRCh38
5342474 c.471G>A p.Q157Q Chr.1: 46260605-46260605 on Build GRCh38
459127 c.460C>T p.R154W Chr.1: 46260594-46260594 on Build GRCh38
5904121 c.418C>T p.P140S Chr.1: 46260552-46260552 on Build GRCh38
6360452 c.461G>T p.R154L Chr.1: 46260595-46260595 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1008084545 Chr.1: 46260580-46260580 on Build GRCh38
rs147070253 Chr.1: 46260563-46260563 on Build GRCh38
rs763307926 Chr.1: 46260529-46260529 on Build GRCh38
rs28363217 Chr.1: 46260483-46260483 on Build GRCh38
rs747012334 Chr.1: 46260507-46260507 on Build GRCh38
rs780246007 Chr.1: 46260601-46260601 on Build GRCh38
rs775770168 Chr.1: 46260528-46260528 on Build GRCh38
rs200522718 Chr.1: 46260494-46260494 on Build GRCh38
rs568709743 Chr.1: 46260445-46260445 on Build GRCh38
rs762275372 Chr.1: 46260550-46260550 on Build GRCh38
rs148273228 Chr.1: 46260607-46260607 on Build GRCh38
rs376801907 Chr.1: 46260596-46260596 on Build GRCh38
rs746174568 Chr.1: 46260488-46260488 on Build GRCh38
rs202208887 Chr.1: 46260551-46260551 on Build GRCh38
rs149141765 Chr.1: 46260594-46260594 on Build GRCh38
rs754609210 Chr.1: 46260565-46260565 on Build GRCh38
rs964003694 Chr.1: 46260597-46260597 on Build GRCh38
rs199693607 Chr.1: 46260617-46260617 on Build GRCh38
rs769944658 Chr.1: 46260523-46260523 on Build GRCh38
rs370579299 Chr.1: 46260475-46260475 on Build GRCh38
rs374634207 Chr.1: 46260514-46260514 on Build GRCh38
rs777770167 Chr.1: 46260581-46260581 on Build GRCh38
rs764507127 Chr.1: 46260530-46260530 on Build GRCh38
rs752474512 Chr.1: 46260574-46260574 on Build GRCh38
rs975812833 Chr.1: 46260603-46260603 on Build GRCh38
rs776626477 Chr.1: 46260515-46260515 on Build GRCh38
rs757184891 Chr.1: 46260588-46260588 on Build GRCh38
rs754488431 Chr.1: 46260554-46260554 on Build GRCh38
rs371935755 Chr.1: 46260595-46260595 on Build GRCh38
rs774823254 Chr.1: 46260532-46260532 on Build GRCh38
rs531772499 Chr.1: 46260430-46260432 on Build GRCh38
rs890296809 Chr.1: 46260600-46260600 on Build GRCh38
rs535998793 Chr.1: 46260462-46260462 on Build GRCh38
rs2295466 Chr.1: 46260585-46260585 on Build GRCh38
rs764812486 Chr.1: 46260567-46260567 on Build GRCh38
rs772504382 Chr.1: 46260604-46260605 on Build GRCh38
rs778354255 Chr.1: 46260557-46260557 on Build GRCh38

View Full Product Details