Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 84064786-84065039 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPS6KA6
Entrez Gene ID
27330
Gene Name
ribosomal protein S6 kinase A6
Gene Aliases
PP90RSK4, RSK4
UniGene
Hs.234478
Gene Chromosome Location
Chr.X: 84058346-84187935 on Build GRCh38
Transcripts
NM_014496.4 XM_011530917.2 XM_011530918.2 XM_011530919.2 XM_011530920.2 XM_017029423.1 XM_017029424.1 XM_017029425.1 XM_017029426.1

SNP Detail

SNP ID Chromosome Location
rs1006690228 Chr.X: 84064840-84064840 on Build GRCh38
rs748795418 Chr.X: 84064889-84064892 on Build GRCh38
rs958542234 Chr.X: 84064938-84064938 on Build GRCh38
rs757201273 Chr.X: 84064950-84064951 on Build GRCh38
rs925531577 Chr.X: 84064919-84064919 on Build GRCh38
rs1031413198 Chr.X: 84064833-84064833 on Build GRCh38
rs369916805 Chr.X: 84064941-84064941 on Build GRCh38
rs775301300 Chr.X: 84064874-84064874 on Build GRCh38
rs535435468 Chr.X: 84064844-84064844 on Build GRCh38
rs781127325 Chr.X: 84064834-84064834 on Build GRCh38
rs975070000 Chr.X: 84064935-84064935 on Build GRCh38
rs369351467 Chr.X: 84064942-84064942 on Build GRCh38
rs370645295 Chr.X: 84064961-84064961 on Build GRCh38
rs762731263 Chr.X: 84064895-84064895 on Build GRCh38
rs966682708 Chr.X: 84064894-84064894 on Build GRCh38
rs981806270 Chr.X: 84064954-84064954 on Build GRCh38
rs951974052 Chr.X: 84064905-84064905 on Build GRCh38
rs6616890 Chr.X: 84065009-84065009 on Build GRCh38
rs955879790 Chr.X: 84064843-84064844 on Build GRCh38
rs781616040 Chr.X: 84064928-84064928 on Build GRCh38
rs888372025 Chr.X: 84064821-84064821 on Build GRCh38
rs766104053 Chr.X: 84064899-84064899 on Build GRCh38
rs377421367 Chr.X: 84064949-84064949 on Build GRCh38
rs779053870 Chr.X: 84064892-84064892 on Build GRCh38
rs769545082 Chr.X: 84064870-84064870 on Build GRCh38

View Full Product Details